Skip to Content
Merck
All Photos(1)

Key Documents

93834

Sigma-Aldrich

2-Chloro-4-nitrophenyl-α-D-maltotrioside

chromogenic, ≥95% (HPLC), powder or crystals

Synonym(s):

2-chloro-4-nitrophenyl O-α-D-glucopyranosyl-(1→4)-O-α-D-glucopyranosyl-(1→4)-α-D-glucopyranoside, CNP-G3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H34ClNO18
CAS Number:
Molecular Weight:
659.98
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32

product name

2-Chloro-4-nitrophenyl-α-D-maltotrioside, ≥95% (HPLC)

Quality Level

Assay

≥95% (HPLC)

form

powder or crystals

storage temp.

−20°C

SMILES string

OC[C@H]1O[C@H](O[C@H]2[C@H](O)[C@@H](O)[C@H](O[C@@H]2CO)O[C@H]3[C@H](O)[C@@H](O)[C@H](O[C@@H]3CO)Oc4ccc(cc4Cl)[N+]([O-])=O)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C24H34ClNO18/c25-8-3-7(26(37)38)1-2-9(8)39-22-18(35)15(32)20(11(5-28)41-22)44-24-19(36)16(33)21(12(6-29)42-24)43-23-17(34)14(31)13(30)10(4-27)40-23/h1-3,10-24,27-36H,4-6H2/t10-,11-,12-,13-,14+,15-,16-,17-,18-,19-,20-,21-,22+,23-,24-/m1/s1

InChI key

KMYYNUOXSFGLNX-XFNLHOCBSA-N

Application

2-Chloro-4-nitrophenyl-α-D-maltotrioside has been used as a substrate in assays to determine the enzymatic activity of α-amylase.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sergio Alcalá-Alcalá et al.
International journal of pharmaceutics, 489(1-2), 139-147 (2015-05-06)
This work presents an evaluation of the adsorption/infiltration process in relation to the loading of a model protein, α-amylase, into an assembled biodegradable polymeric system, free of organic solvents and made up of poly(D,L-lactide-co-glycolide) acid (PLGA). Systems were assembled in
T Suganuma et al.
Carbohydrate research, 303(2), 219-227 (1997-11-14)
The degradation mechanism of a synthetic substrate, 2-chloro-4-nitrophenyl alpha-maltotrioside (CNP-G3), by human salivary alpha-amylase (HSA) was investigated by kinetic and product analyses. It was observed that the enzyme attacked the various CNP-maltooligosaccharides (CNP-G3 to CNP-G6) releasing free CNP. Addition of
K Lorentz et al.
Clinical chemistry and laboratory medicine, 37(11-12), 1053-1062 (2000-03-22)
We present the adaptation of an IFCC method for alpha-amylase using 2-chloro-4-nitro-phenyl-alpha-D-maltotrio-side as substrate (1) suited for routine work at 37 degrees C. In the assay, a constant proportion of substrate, i. e. 92%, is directly converted to 2-chloro-4-nitrophenol and
alpha-Amylase and alpha-glucosidase inhibitory saponins from Polyscias fruticosa leaves
Hanh TTH, et al.
Journal of Chemistry (2016)
Kinetic determination of amylase on microtiter plates: an improved substrate.
J Laine et al.
Pancreas, 13(2), 217-217 (1996-08-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service