Skip to Content
Merck
All Photos(1)

Key Documents

93815

Sigma-Aldrich

Rhamnazin

≥99% (HPLC)

Synonym(s):

3′,7-Di-O-methylquercetin, 3′,7-Dimethylquercetin, 3,4′,5-Trihydroxy-3′,7-dimethoxyflavone, Quercetin 3′,7-dimethyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H14O7
CAS Number:
Molecular Weight:
330.29
Beilstein:
338203
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥99% (HPLC)

form

powder

SMILES string

COc1cc(O)c2C(=O)C(O)=C(Oc2c1)c3ccc(O)c(OC)c3

InChI

1S/C17H14O7/c1-22-9-6-11(19)14-13(7-9)24-17(16(21)15(14)20)8-3-4-10(18)12(5-8)23-2/h3-7,18-19,21H,1-2H3

InChI key

MYMGKIQXYXSRIJ-UHFFFAOYSA-N

Application

Rhamnazin may be used as a reference material in analysis of plant flavonoids. Rhamnazin and its glycosides may be used to study their potentials as DNA topoisomerase I inhibitors/poisons and as anitoxidants.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B S Yun et al.
Archives of pharmacal research, 23(2), 147-150 (2000-06-03)
Twelve compounds with lipid peroxidation inhibitory activity were isolated from the stem bark of E. globulus. Their structures were assigned as a new aromatic monoterpene (1) and eleven known compounds, pinoresinol (2), vomifoliol (3), 3,4,5-trimethoxyphenol 1-O-beta-D-(6'-O-galloyl)glucopyranoside (4), methyl gallate (5)
M López-Lázaro et al.
Journal of enzyme inhibition and medicinal chemistry, 17(1), 25-29 (2002-10-09)
The therapeutic anticancer potential of flavonoids shown by recent research needs a greater understanding of these compounds. They are antioxidants and antimutagenic agents that can inhibit tumor promotion and transformation and can modify the activity of a large number of
Yao Yu et al.
Biochemical and biophysical research communications, 458(4), 913-919 (2015-02-24)
Anti-angiogenesis targeting vascular endothelial growth factor receptor 2 (VEGFR2) has emerged as an important tool for cancer therapy. The identification of new drugs from natural products has a long and successful history. In this study, we described a novel VEGFR2
M López-Lázaro et al.
Zeitschrift fur Naturforschung. C, Journal of biosciences, 55(11-12), 898-902 (2001-02-24)
Flavonoids are secondary plant metabolites whose anticancer properties are actually being studied from an epidemiological and pharmacological point of view. They are believed to be implicated in the lower risk of some forms of cancer observed in Asian countries, due
Mervat M A El-Gendy et al.
Applied biochemistry and biotechnology, 150(1), 85-96 (2008-06-14)
In our searching program for bioactive secondary metabolites from marine Streptomycetes, three microbial benzopyrone derivatives (1-3), 7-methylcoumarin (1) and two flavonoides, rhamnazin (2) and cirsimaritin (3), were obtained during the working up of the ethyl acetate fraction of a marine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service