Skip to Content
Merck
All Photos(1)

Key Documents

86826

Sigma-Aldrich

5-Carboxyfluorescein

BioReagent, suitable for fluorescence, ≥95% (HPLC)

Synonym(s):

5-FAM

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H12O7
CAS Number:
Molecular Weight:
376.32
Beilstein:
57037
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.32

product line

BioReagent

Quality Level

Assay

≥95% (HPLC)

form

powder

solubility

DMF: soluble
DMSO: soluble
H2O: soluble

fluorescence

λex 492 nm; λem 517 nm in 0.1 M Tris pH 8.0

suitability

suitable for fluorescence

SMILES string

OC(=O)c1ccc2c(c1)C(=O)OC23c4ccc(O)cc4Oc5cc(O)ccc35

InChI

1S/C21H12O7/c22-11-2-5-15-17(8-11)27-18-9-12(23)3-6-16(18)21(15)14-4-1-10(19(24)25)7-13(14)20(26)28-21/h1-9,22-23H,(H,24,25)

InChI key

NJYVEMPWNAYQQN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

5-Carboxyfluorescein, also known as 5-FAM, is a green fluorescent product. It is derived from cleavage of 5-Carboxyfluorescein diacetate by intracellular esterases. In viable cells, the fluorescent molecule is impermeable to the cellular membrane.

Application

5-Carboxyfluorescein has been used as a payload to study leakage phenomena on sesame (Sesamum indicum L.) and coconut (Cocos nucifera L.) liposomes.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Derivative of fluorescein giving stable derivative upon conjugation with biopolymers

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Detection of Bacteria, Viruses, Parasites and Fungi (2010)
Study on leakage of sesame (Sesamum indicum L.) and coconut (Cocos nucifera L.) liposomes
Hudiyanti D, et al.
Orient. J. Chem., 31, 435-439 (2015)
The loss-of-allele assay for ES cell screening and mouse genotyping
Methods in Enzymology, 476, 295-307 (2010)
Intracellular pH and its Measurement (1989)
Nicolas Corradi et al.
Molecular genetics and genomics : MGG, 280(6), 565-574 (2008-09-27)
Microsporidia are a group of intracellular parasites characterized by highly reduced and compact genomes. The presence of a high gene density had several consequences for microsporidian genomes, including a high frequency of overlap between transcripts of adjacent genes. This phenomenon

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service