71679
Sodium bitartrate monohydrate
BioUltra, ≥99.0% (T)
Synonym(s):
L-(+)-Tartaric acid monosodium salt, Sodium hydrogen tartrate monohydrate
About This Item
Recommended Products
product line
BioUltra
Quality Level
Assay
≥99.0% (T)
form
powder or crystals
optical activity
[α]20/D +22±1°, c = 1% in H2O
impurities
insoluble matter, passes filter test
pH
2.5-4.0 (25 °C, 0.1 M in H2O)
solubility
H2O: 0.1 M at 20 °C, clear, colorless
anion traces
chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤50 mg/kg
cation traces
Al: ≤5 mg/kg
As: ≤0.1 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤50 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg
λ
0.1 M in H2O
UV absorption
λ: 260 nm Amax: 0.01
λ: 280 nm Amax: 0.01
SMILES string
[Na+].[H]O[H].OC(C(O)C([O-])=O)C(O)=O
InChI
1S/C4H6O6.Na.H2O/c5-1(3(7)8)2(6)4(9)10;;/h1-2,5-6H,(H,7,8)(H,9,10);;1H2/q;+1;/p-1
InChI key
LLVQEXSQFBTIRD-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
Related Categories
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service