Skip to Content
Merck
All Photos(1)

Key Documents

71162

Sigma-Aldrich

Naringin

≥95% (HPLC)

Synonym(s):

4′,5,7-Trihydroxyflavanone 7-rhamnoglucoside, Naringenin 7-neohesperidoside, Naringenine-7-rhamnosidoglucoside, Naringoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H32O14
CAS Number:
Molecular Weight:
580.53
Beilstein:
102012
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.47

Quality Level

Assay

≥95% (HPLC)

form

powder

optical activity

[α]20/D −80±10°, c = 1% in ethanol

mp

83  °C ((181 °F ))

solubility

ethanol: o.1 g/10 mL, clear to hazy, colorless to light greenish-yellow

SMILES string

C[C@@H]1O[C@@H](O[C@@H]2[C@@H](O)[C@H](O)[C@@H](CO)O[C@H]2Oc3cc(O)c4C(=O)CC(Oc4c3)c5ccc(O)cc5)[C@H](O)[C@H](O)[C@H]1O

InChI

1S/C27H32O14/c1-10-20(32)22(34)24(36)26(37-10)41-25-23(35)21(33)18(9-28)40-27(25)38-13-6-14(30)19-15(31)8-16(39-17(19)7-13)11-2-4-12(29)5-3-11/h2-7,10,16,18,20-30,32-36H,8-9H2,1H3/t10-,16?,18+,20-,21+,22+,23-,24+,25+,26-,27+/m0/s1

InChI key

DFPMSGMNTNDNHN-JJLSSNRUSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Naringin has been used:
  • as a bitter tastant to compare the behavioral response of the Drosophila larva and adult(2)
  • to study its anti-inflammatory property and to determine its effect on nucleus pulposus (NP) cells(3)
  • to determine its effect on bone metabolism like osteogenic differentiation, inhibition of osteoclast formation(4)

Biochem/physiol Actions

Naringin can interact with signaling molecules and modulates signaling pathways. It possesses anti-inflammatory and anti-cancer activities. It also exhibits pharmacological effects on oxidative stress, bone regeneration, genetic damage, metabolic syndrome and central nervous system (CNS) diseases.
Naringin, a flavanoid in grapefruit and other citrus fruits, potently inhibits intestinal organic anion-transporting polypeptide 1A2 (OATP1A2). Grapefruit juice thereby reduces bioavailability of many pharmacological agents taken at the same time.

Other Notes

To gain a comprehensive understanding of our extensive range of Disaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marcelo Silva et al.
Industrial & engineering chemistry research, 57(28), 9210-9221 (2018-10-03)
Liquid-liquid extraction (LLE) can be an effective strategy for the purification of polyphenols from a fermentation broth. However, solvents need to be chosen to ensure high extraction capacity and selectivity. For that purpose, a systematic study is here presented, where
Therapeutic potential of naringin: an overview
Chen R, et al.
Pharmaceutical biology, 54(12), 3203-3210 (2016)
Luís F Mendes et al.
Scientific reports, 9(1), 14906-14906 (2019-10-19)
The ability of flavonoids to attenuate macrophage pro-inflammatory activity and to promote macrophage-mediated resolution of inflammation is still poorly understood at the biochemical level. In this study, we have employed NMR metabolomics to assess how therapeutically promising flavonoids (quercetin, naringenin
Susana Ferreyra et al.
Food chemistry, 338, 128030-128030 (2020-09-16)
A liquid chromatography method coupling diode-array and fluorescence detectors (DAD and FLD, respectively) has been developed for the simultaneous quantification of 32 phenolic compounds (PCs) in winemaking products. With the combination of both detectors it was possible to determine phenolic
Jiukai Zhang et al.
Food chemistry, 135(3), 1471-1478 (2012-09-08)
Huyou (Citrus changshanensis) is rich in naringin and neohesperidin, which are natural flavanone glycosides with a range of biological activities. Among the different fruit parts, i.e. flavedo, albedo, segment membrane (SM), and juice sacs (JS), albedo showed the highest contents

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service