Skip to Content
Merck
All Photos(1)

Key Documents

56822

Sigma-Aldrich

Immersion oil

for microscopy

Synonym(s):

immersion medium for light microscopy

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12171500
NACRES:
NA.47

biological source

synthetic

Quality Level

grade

for microscopy

form

liquid

refractive index

n20/D 1.516

viscosity

100-120 mPa.s(20 °C)

density

1.025 g/mL at 20 °C

application(s)

hematology
histology

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

General description

Immersion oil is a clear, viscous liquid with optimized refractive properties, specifically modified to closely approximate the refractive index (RI) of glass (ne = 1.5). It is used in conjunction with an objective lens to enhance the resolving power.

Application

Immersion oil is used for high-resolution (1000X) light microscopy work in conjunction with an oil immersion objective lens to optimize microscopic examinations of histological, cytological, hematological, and bacterial specimen material after it has been fixed, embedded, stained, or counterstained, and mounted.

Principle

Immersion oil is applied dropwise to stained and mounted or non-mounted specimen material to form a clear film between the specimen and the microscope lens to eliminate the deflection of incident light and thus substantially enhance the optical efficiency of the lens.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Theis Sommer et al.
Scientific reports, 8(1), 13104-13104 (2018-09-01)
The catalytic mechanism of the cyclic amidohydrolase isatin hydrolase depends on a catalytically active manganese in the substrate-binding pocket. The Mn2+ ion is bound by a motif also present in other metal dependent hydrolases like the bacterial kynurenine formamidase. The
P L Appleton et al.
Journal of microscopy, 234(2), 196-204 (2009-04-29)
Visualizing overall tissue architecture in three dimensions is fundamental for validating and integrating biochemical, cell biological and visual data from less complex systems such as cultured cells. Here, we describe a method to generate high-resolution three-dimensional image data of intact
Thomas P Burghardt et al.
Applied optics, 48(32), 6120-6131 (2009-11-12)
Total internal reflection fluorescence (TIRF) microscopy uses the evanescent field on the aqueous side of a glass/aqueous interface to selectively illuminate fluorophores within approximately 100 nm of the interface. Applications of the method include epi-illumination TIRF, where the exciting light

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service