52583
Lipase B Candida antarctica immobilized on Immobead 150, recombinant from yeast
≥2000 U/g
Synonym(s):
Candida antarctica Lipase
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352204
NACRES:
NA.54
Recommended Products
recombinant
expressed in yeast
Quality Level
form
beads
specific activity
≥2000 U/g
mol wt
33 kDa
storage temp.
2-8°C
General description
Lipase B or triacylglycerol ester hydrolases belongs to the class of hydrolases. Lipase B from Candida antarctica (CALB) possesses 317 amino acids and a molecular weight of 33 kDa. CALB has a catalytic triad and an open solvent accessible active site. The catalytic triad is usually Ser-His-Asp/Glu which is found in the carboxy terminal of parallel β sheet.
Application
Lipase B Candida antarctica immobilized on Immobead 150, recombinant from yeast has been used:
- in esterification reaction of lauric acid with n-butanol in a biphasic solvent system
- in hydrolysis of fish oil triglycerides
- in screening of enzymes for Morita–Baylis–Hillman (MBH) reaction
Lipases are used industrially for the resolution of chiral compounds and the transesterification production of biodiesel.
Biochem/physiol Actions
Lipase B from Candida antarctica has been shown to be an effective catalyst for the synthesis of esters of ethyl D-glucopyranoside from fatty acids larger than octanoic acid. It has also been found to catalyze a wide variety of organic reactions including many different regio- and enantio-selective syntheses.
Lipases B from Candida antarctica (CALB) is a versatile catalyst for biotransformation reactions. In aqueous media, CALB behaves like an esterase rather than a lipase.
Lipases catalyze the hydrolysis of triacylglycerols into glycerol and free fatty acids.
Unit Definition
1 U corresponds to the amount of enzyme which liberates 1 μmol butyric acid per minute at pH 7.5 and 40°C (tributyrin, Cat. No. 91010, as substrate)
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Thermodynamic study of hydrolysis and esterification reactions with immobilized lipases
Sharma A, et al.
European International Journal of Science and Technology, 4, 128-136 (2015)
Study of reaction parameters and kinetics of esterification of lauric acid with butanol by immobilized Candida antarctica lipase
Shankar S, et al.
Indian Journal of Biochemistry & Biophysics, 50, 570-576 (2013)
Fatty Acid Specificity in Lipase-Catalyzed Synthesis of Glucoside Esters
O. Kirk et al.
Biocatalysis and Biotransformation, 6, 127-134 (1992)
Kinetics of acyl transfer reactions in organic media catalysed by Candida antarctica lipase B
Martinelle M and Hult K
Biochimica et Biophysica Acta, Protein Structure and Molecular Enzymology, 1251(2), 191-197 (1995)
The sequence, crystal structure determination and refinement of two crystal forms of lipase B from Candida antarctica
Uppenberg J, et al.
Structure, 2(4), 293-308 (1994)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service