Skip to Content
Merck
All Photos(1)

Key Documents

52444

Sigma-Aldrich

Violaxanthin

≥90.0% (HPLC), solid

Synonym(s):

(3S,3′S,5R,5′R,6S,6′S)-5,6:5′,6′-Diepoxy-5,5′,6,6′-tetrahydro-β,β-carotene-3,3′-diol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H56O4
CAS Number:
Molecular Weight:
600.87
Beilstein:
101269
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.32

Product Name

Violaxanthin, ≥90.0% (HPLC)

Quality Level

Assay

≥90.0% (HPLC)

form

solid

λ

in ethanol

UV absorption

λ: 438-442 nm Amax

storage temp.

−20°C

SMILES string

CC(/C=C/[C@@]12[C@](C)(O2)C[C@@H](O)CC1(C)C)=C\C=C\C(C)=C\C=C\C=C(C)\C=C\C=C(\C=C\[C@]34C(C)(C)C[C@H](O)C[C@]3(O4)C)C

InChI

1S/C40H56O4/c1-29(17-13-19-31(3)21-23-39-35(5,6)25-33(41)27-37(39,9)43-39)15-11-12-16-30(2)18-14-20-32(4)22-24-40-36(7,8)26-34(42)28-38(40,10)44-40/h11-24,33-34,41-42H,25-28H2,1-10H3/b12-11+,17-13+,18-14+,23-21+,24-22+,29-15+,30-16+,31-19+,32-20+/t33-,34-,37+,38+,39-,40-/m0/s1

InChI key

SZCBXWMUOPQSOX-WVJDLNGLSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Violaxanthin is an epoxidized carotenoid obtained from plants. It is an orange-colored natural xanthophyll pigment.

Application

Violaxanthin has been used as a standard/control in high-pressure liquid chromatography (HPLC) analysis of pigment extracts to confirm the high-level astaxanthin accumulation suspected from the visual phenotype of Nt-AXT plants.

Biochem/physiol Actions

Violaxanthin is a carotene epoxide that is a precursor to capsanthin. It is one of three xanthophylls involved in the evolution of plastids of green plants (oxygen evolution) and participating in photo-induced interconversions known as the violaxanthin cycle. The cleavage of 9-cis-epoxycarotenoids (violaxanthin) to xanthoxin is the key regulatory step of abscisic acid biosynthesis.
Violaxanthin is a powerful antioxidant and a vital precursor to important compounds like fucoxanthin and β-damascenone. It is a non-photochemical fluorescence quenching structural component of the xanthophyll cycle that shields the photosynthetic system from excessive light. The 2,2′-azinobis-3-ethylbenzothiazoline-6-sulfonic acid (ABTS+) radical scavenging action of violaxanthin extracted from water spinach/water morning glory is strong, and it also inhibits lipid peroxidation and red blood cell hemolysis. Hence, it has the potential to be used in a wide range of medicinal and health products.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Photochemistry of Carotenoids (2006)
K K Niyogi et al.
The Plant cell, 10(7), 1121-1134 (1998-07-21)
A conserved regulatory mechanism protects plants against the potentially damaging effects of excessive light. Nearly all photosynthetic eukaryotes are able to dissipate excess absorbed light energy in a process that involves xanthophyll pigments. To dissect the role of xanthophylls in
Yinghong Lu et al.
Current biology : CB, 27(19), 3034-3041 (2017-09-26)
Transgene expression from the plastid (chloroplast) genome provides unique advantages, including high levels of foreign protein accumulation, convenient transgene stacking in operons, and increased biosafety due to exclusion of plastids from pollen transmission [1, 2]. However, applications in biotechnology and
The spectral determination of chlorophylls a and b, as well as total carotenoids, using various solvents with spectrophotometers of different resolution.
Wellburn, A.R.
Journal of Plant Physiology, 144, 307-313 (1994)
Daniel Riebe et al.
Sensors (Basel, Switzerland), 19(23) (2019-12-05)
The lack of soil data, which are relevant, reliable, affordable, immediately available, and sufficiently detailed, is still a significant challenge in precision agriculture. A promising technology for the spatial assessment of the distribution of chemical elements within fields, without sample

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service