Skip to Content
Merck
All Photos(1)

Key Documents

51120

Sigma-Aldrich

Guanosine 5′-triphosphate sodium salt hydrate

≥90% (HPLC)

Synonym(s):

GTP, GTP-Na⁺ hydrate, 5′-GTP-Na2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16N5O14P3 · xNa+ · yH2O
CAS Number:
Molecular Weight:
523.18 (anhydrous free acid basis)
Beilstein:
4113439
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

Quality Level

Assay

≥90% (HPLC)

form

powder

optical activity

[α]20/D −24±2°, c = 1% in 0.5 M Na2HPO4

impurities

≤3 mol/mol water (NMR)

shipped in

wet ice

storage temp.

−20°C

SMILES string

O.O.[Na+].[Na+].NC1=Nc2c(ncn2[C@@H]3O[C@H](COP(O)(=O)OP([O-])(=O)OP(O)([O-])=O)[C@@H](O)[C@H]3O)C(=O)N1

InChI

1S/C10H16N5O14P3.2Na.2H2O/c11-10-13-7-4(8(18)14-10)12-2-15(7)9-6(17)5(16)3(27-9)1-26-31(22,23)29-32(24,25)28-30(19,20)21;;;;/h2-3,5-6,9,16-17H,1H2,(H,22,23)(H,24,25)(H2,19,20,21)(H3,11,13,14,18);;;2*1H2/q;2*+1;;/p-2/t3-,5-,6-,9-;;;;/m1..../s1

InChI key

HPLNHYUZFVUXFJ-ZVQJTLEUSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Guanosine 5′-triphosphate (GTP) is a purine trinucleotide. GTP is a substrate for a wide range of G-protein coupled receptor (GPCR) GTPases, as well as cell signaling and cycling-associated guanine nucleotide exchange factors (GEF). See product G3776 for RNA polymerase transcription and DNA or RNA biosynthesis applications.

Application

Guanosine 5′-triphosphate sodium salt hydrate has been used:
  • as a component of the internal solution for astrocyte patch-clamp to monitor astrocyte calcium activity from mouse amygdala slices.
  • in a solution for channel current recording in cell lines using a voltage-clamp technique.
  • to prepare internal recording solution for injectoporation to deliver genes to mechanosensory hair cells.
  • promoting the formation of G interface-mediated homodimers of human septin during the analysis of its oligomerization state by analytical size exclusion chromatography (SEC).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hiroaki Kajiho et al.
The Journal of biological chemistry, 286(27), 24364-24373 (2011-05-19)
The small GTPase Rab5, which cycles between GDP-bound inactive and GTP-bound active forms, plays essential roles in membrane budding and trafficking in the early endocytic pathway. Rab5 is activated by various vacuolar protein sorting 9 (VPS9) domain-containing guanine nucleotide exchange
Gyu-Sang Hong et al.
Neuron, 94(2), 271-273 (2017-04-21)
Mechanosensation is essential for various physiological processes, and it is mediated by mechanotransduction channels. Recently, we reported that TMEM150C/Tentonin 3 (TTN3) confers mechanically activated currents with slow inactivation kinetics in several cell types, including dorsal root ganglion neurons (Hong et al.
Lukas Jakob Neukomm et al.
Development (Cambridge, England), 138(10), 2003-2014 (2011-04-15)
Multicellular organisms use programmed cell death to eliminate unwanted or potentially harmful cells. Improper cell corpse removal can lead to autoimmune diseases. The development of interventional therapies that increase engulfment activity could represent an attractive approach to treat such diseases.
Angel Baudon et al.
STAR protocols, 3(1), 101159-101159 (2022-02-25)
Astrocytes are glial cells that exhibit calcium signaling-mediated activity. Here, we present a protocol to monitor and manipulate astrocyte calcium activity from mouse amygdala slices. In the first part of this protocol, we describe the procedure of astrocyte calcium imaging.
J Spychala et al.
The International journal of biochemistry, 23(10), 1155-1159 (1991-01-01)
1. Phosphocellulose column chromatography under double gradient conditions (phosphate and KCl) revealed two forms of AMP deaminase in rat heart and brain and a single form in the liver and skeletal muscle. 2. Kinetically all purified AMP deaminases were classified

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service