Skip to Content
Merck
All Photos(1)

Key Documents

5085

Sigma-Aldrich

E-Cadherin human

recombinant, expressed in E. coli, 0.5 mg protein/mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.75

biological source

human

recombinant

expressed in E. coli

description

0.1 mg recombinant human E-Cadherin in 20 mM Tris-HCl buffer, containing NaCl, KCl, EDTA, L-arginine, DTT and glycerol.

sterility

Filtered sterilized solution

Assay

≥90% (SDS-PAGE)

form

liquid

packaging

pkg of 100 μg

concentration

0.5 mg protein/mL

technique(s)

cell culture | mammalian: suitable

accession no.

NP_004351.1

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... CDH1(999)

Application

E-Cadherin has been used as coating matrix protein for 1) maintaining long-term ES or iPS cell culture then combine with proper ES cell culture media, 2) as a coating matrix material for 11R tag recombinant TF intracellular delivery for protein derived iPS protocol with extremely low-level non-specific interaction and 3) as a native antigen for antibody production.

Use these recommendations as guidelines to determine the optimal coating conditions for your culture system.
1. Thaw E-Cadherin and dilute to desired concentration using serum-free medium or PBS. The final solution should be sufficiently dilute so that the volume added covers the surface evenly.
Note: Use 1 ml PBS per well in a 6-well plate.
2. Add 5 - 10 μg protein to each well and incubate at 2 to 10 °C overnight.
3. After incubation, aspirate remaining material.
4. Plates are ready for use. They may also be stored at 2-8 °C damp or air dried if sterility is maintained.

Sequence

MDWVIPPISCPENEKGPFPKNLVQIKSNKDKEGKVFYSITGQGADTPPVGVFIIERETGWLKVTEPLDRERIATYTLFSHAVSSNGNAVEDPMEILITVTDQNDNKPEFTQEVFKGSVMEGALPGTSVMEVTATDADDDVNTYNAAIAYTILSQDPELPDKNMFTINRNTGVISVVTTGLDRESFPTYTLVVQAADLQGEGLSTTATAVITVTDTNDNPPIFNPTTYKGQVPENEANVVITTLKVTDADAPNTPAWEAVYTILNDDGGQFVVTTNPVNNDGILKTAKGLDFEAKQQYILHVAVTNVVPFEVSLTTSTATVTVDVLDVNEAPIFVPPEKRVEVSEDFGVGQEITSYTAQEPDTFMEQKITYRIWRDTANWLEINPDTGAISTRAELDREDFEHVKNSTYTALIIATDNGSPVATGTGTLLLILSDVNDNAPIPEPRTIFFCERNPKPQVINIIDADLPPNTSPFTAELTHGASANWTIQYNDPTQESIILKPKMALEVGDYKINLKLMDNQNKDQVTTLEVSVCDCEGAAGVCRKAQPVEAGLQIP

Preparation Note

Recombinant human E-Cadherin gene (155-710 aa Fragment) was constructed with codon optimization and expressed in non-fusion protein form in E. coli as inclusion bodies. The final product was refolded using a unique "temperature shift inclusion body refolding" technology and chromatographically purified.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tomomi Izumikawa et al.
Scientific reports, 4, 3701-3701 (2014-01-16)
Chondroitin sulfate (CS) proteoglycans are present on the surfaces of virtually all cells and in the extracellular matrix and are required for cytokinesis at early developmental stages. Studies have shown that heparan sulfate (HS) is essential for maintaining mouse embryonic
Maryam Salimi et al.
The Journal of experimental medicine, 210(13), 2939-2950 (2013-12-11)
Type 2 innate lymphoid cells (ILC2s, nuocytes, NHC) require RORA and GATA3 for their development. We show that human ILC2s express skin homing receptors and infiltrate the skin after allergen challenge, where they produce the type 2 cytokines IL-5 and
Evgeny A Moskalev et al.
BMC cancer, 12, 213-213 (2012-06-08)
The Wnt/β-catenin signalling is aberrantly activated in primary B cell chronic lymphocytic leukaemia (CLL). Epigenetic silencing of pathway inhibitor genes may be a mechanism for its activation. In this study, we investigated systematically and quantitatively the methylation status of 12
J O Humtsoe et al.
Oncogene, 29(8), 1214-1226 (2009-11-26)
Tumor cells are capable of surviving loss of nutrients and anchorage in hostile microenvironments. Under these conditions, adapting to specific signaling pathways may shift the balance between growth and cellular dormancy. Here, we report a mechanism by which epidermal growth
Yue Xu et al.
Proceedings of the National Academy of Sciences of the United States of America, 107(18), 8129-8134 (2010-04-22)
Using a high-throughput chemical screen, we identified two small molecules that enhance the survival of human embryonic stem cells (hESCs). By characterizing their mechanisms of action, we discovered an essential role of E-cadherin signaling for ESC survival. Specifically, we showed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service