Skip to Content
Merck
All Photos(1)

Key Documents

48880

Sigma-Aldrich

Gibberellic acid

90% gibberellin A3 basis (HPLC)

Synonym(s):

GA3, Gibberellin A3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H22O6
CAS Number:
Molecular Weight:
346.37
Beilstein:
54346
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

90% gibberellin A3 basis (HPLC)

optical activity

[α]20/D +78±5°, c = 2% in methanol

SMILES string

C[C@]12[C@@H](O)C=C[C@@]3(OC1=O)[C@@H]4CC[C@]5(O)C[C@]4(CC5=C)[C@H]([C@H]23)C(O)=O

InChI

1S/C19H22O6/c1-9-7-17-8-18(9,24)5-3-10(17)19-6-4-11(20)16(2,15(23)25-19)13(19)12(17)14(21)22/h4,6,10-13,20,24H,1,3,5,7-8H2,2H3,(H,21,22)/t10-,11+,12-,13-,16-,17+,18+,19-/m1/s1

InChI key

IXORZMNAPKEEDV-OBDJNFEBSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Gibberellic acid is an endogenous plant growth regulator

Application

Gibberellic acid has been used to study mechanical properties with cellular resolution in the light-grown hypocotyl of Arabidopsis thaliana.

Biochem/physiol Actions

Phytohormone that controls important aspects of plant growth: germination, elongation, and flowering. The effects are mediated by inducing degradation of the DELLA repressor protein, and probably other minor signalling pathways.

Other Notes

Structure-activity studies

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

An automated confocal micro-extensometer enables in vivo quantification of mechanical properties with cellular resolution
Robinson S, et al.
Plant Cell, 29(12), 2959-2973 (2017)
E.P. Serebryakov et al.
Phytochemistry, 23, 1847-1847 (1984)
Gokhan Kir et al.
Plant physiology, 169(1), 826-839 (2015-07-15)
Brassinosteroids (BRs) are plant hormones involved in various growth and developmental processes. The BR signaling system is well established in Arabidopsis (Arabidopsis thaliana) and rice (Oryza sativa) but poorly understood in maize (Zea mays). BRASSINOSTEROID INSENSITIVE1 (BRI1) is a BR
Gemma L Hoyle et al.
Global change biology, 19(5), 1549-1561 (2013-03-19)
Global warming is occurring more rapidly above the treeline than at lower elevations and alpine areas are predicted to experience above average warming in the future. Temperature is a primary factor in stimulating seed germination and regulating changes in seed
Pia Guadalupe Dominguez et al.
Plant physiology, 161(3), 1486-1500 (2013-01-11)
Asr (for ABA, stress, ripening) genes are exclusively found in the genomes of higher plants, and the encoded proteins have been found localized both to the nucleus and cytoplasm. However, before the mechanisms underlying the activity of ASR proteins can

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service