Skip to Content
Merck
All Photos(1)

Key Documents

453M-9

Sigma-Aldrich

p53 (DO7) Mouse Monoclonal Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

100
500

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

DO7, monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (453M-94)
vial of 0.5 mL concentrate (453M-95)
bottle of 1.0 mL predilute (453M-97)
vial of 1.0 mL concentrate (453M-96)
bottle of 7.0 mL predilute (453M-98)

manufacturer/tradename

Cell Marque®

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100-1:500

isotype

IgG2bκ

control

colon carcinoma

shipped in

wet ice

storage temp.

2-8°C

visualization

nuclear

Gene Information

human ... TP53(7157)

General description

Anti-p53 tumor suppressor protein recognizes a 53 kDa hosphoprotein, identified as a p53 suppressor gene product. It reacts with the mutant as well as wild form of p53. Positive nuclear staining with this antibody has been shown to be a negative prognostic factor in breast carcinoma, lung carcinoma, colorectal, and urothelial carcinoma. Anti-p53 positivity has also been used to differentiate uterine serous carcinoma from endometrioid carcinoma as well as to detect intratubular germ cell neoplasia.

Quality


IVD

IVD

IVD

RUO

Linkage

p53 Positive Control Slides, Product No. 453S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: [email protected]

Legal Information

Cell Marque is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Diagnostic Immunohistochemistry: Theranostic and Genomic Applications (2002)
B E Moore et al.
Applied immunohistochemistry & molecular morphology : AIMM, 9(3), 203-206 (2001-09-15)
Intratubular germ cell neoplasia, unclassified (IGCNU) is a precursor of germ cell tumors (GCT) of the testis. In routine histologic sections, neoplastic intratubular germ cells may be very few and easily overlooked. The aim of this study is two-fold: to
F A Mauri et al.
International journal of oncology, 15(6), 1137-1147 (1999-11-24)
Steroid receptor analysis is the only widely accepted prognostic/predictive marker in breast cancer (BC) treatment. In the present study we evaluated the prognostic role of ER/PgR with p53 and Bcl2, in a series of 277 BC (153 pN1/2, 122 pNO
C Midulla et al.
Anticancer research, 19(5B), 4033-4037 (2000-01-11)
The different clinical evolution of breast cancer with similar pathological characteristics prompted the authors to investigate the prognostic significance of different biological markers. Seventy-one primary breast carcinoma specimens obtained by mastectomy or quadrantectomy were examined for the determination of the
D C Quinlan et al.
Cancer research, 52(17), 4828-4831 (1992-09-01)
Mutations in the gene coding for the p53 tumor suppressor protein are common in a variety of human cancers. To assess the role of a putative mutated p53 protein in human lung cancer, a monoclonal antibody recognizing it was used

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service