Skip to Content
Merck
All Photos(1)

Key Documents

42519

Sigma-Aldrich

D-Tagatose 1,6-diphosphate lithium salt

≥90.0% (TLC)

Synonym(s):

TBP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H14O12P2 · xLi+
CAS Number:
Molecular Weight:
340.12 (free acid basis)
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥90.0% (TLC)

storage temp.

2-8°C

SMILES string

O[C@@H]1[C@H](O)C(O)(COP(O)(O)=O)O[C@@H]1COP(O)(O)=O.[Li+]

InChI

1S/C6H14O12P2.Li/c7-4-3(1-16-19(10,11)12)18-6(9,5(4)8)2-17-20(13,14)15;/h3-5,7-9H,1-2H2,(H2,10,11,12)(H2,13,14,15);/q;+1/t3-,4?,5-,6?;/m1./s1

InChI key

GQNKMQYFKBYXBK-WTCPDFSZSA-N

Related Categories

Biochem/physiol Actions

Important metabolite, substrate, activator or inhibitor in carbohydrate utilization pathways like lactose and galactose metabolism.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T D Thomas et al.
Journal of bacteriology, 144(2), 672-682 (1980-11-01)
All of the lactic streptococci examined except Streptococcus lactis ML8 fermented galactose to lactate, formate, acetate, and ethanol. The levels of pyruvate-formate lyase and lactate dehydrogenase were elevated and reduced, respectively, in galactose-grown cells compared with glucose- or lactose-grown cells.
R J van Rooijen et al.
The Journal of biological chemistry, 266(11), 7176-7181 (1991-04-15)
The tagatose 6-phosphate pathway gene cluster (lacABCD) encoding galactose-6-phosphate isomerase, tagatose-6-phosphate kinase, and tagatose-1,6-diphosphate aldolase of Lactococcus lactis subsp. lactis MG1820 has been characterized by cloning, nucleotide sequence analysis, and enzyme assays. Transcription studies showed that the four tagatose 6-phosphate
Amit Mukherjee et al.
Journal of bacteriology, 190(5), 1710-1717 (2007-12-25)
In silico analyses of previously sequenced strains of Escherichia coli O157:H7, EDL933 and Sakai, localized the gene cluster for the utilization of N-acetyl-D-galactosamine (Aga) and D-galactosamine (Gam). This gene cluster encodes the Aga phosphoenolpyruvate:carbohydrate phosphotransferase system (PTS) and other catabolic
E L Rosey et al.
Journal of bacteriology, 173(19), 5992-5998 (1991-10-01)
The nucleotide and deduced amino acid sequences of the lacA and lacB genes of the Staphylococcus aureus lactose operon (lacABCDFEG) are presented. The primary translation products are polypeptides of 142 (Mr = 15,425) and 171 (Mr = 18,953) amino acids
Tobias Fuhrer et al.
Analytical chemistry, 83(18), 7074-7080 (2011-08-13)
Direct injection of samples on high-resolving mass spectrometers is an effective way to maximize analytical throughput and yet allow analyte discrimination in complex samples by mass-to-charge ratio. We present a platform of flow injection electrospray-time-of-flight mass spectrometry to profile small

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service