Skip to Content
Merck
All Photos(1)

Key Documents

33484

Sigma-Aldrich

1,8-Diazafluoren-9-one

Synonym(s):

Cyclopenta[1,2-b:4,3-b’]dipyridin-9-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H6N2O
CAS Number:
Molecular Weight:
182.18
Beilstein:
134499
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.32

Quality Level

mp

229-233 °C

solubility

acetic acid: 10 mg/mL, clear

fluorescence

λex 470 nm; λem 570 nm

storage temp.

2-8°C

SMILES string

O=C1c2ncccc2-c3cccnc13

InChI

1S/C11H6N2O/c14-11-9-7(3-1-5-12-9)8-4-2-6-13-10(8)11/h1-6H

InChI key

FOSUVSBKUIWVKI-UHFFFAOYSA-N

Application

1,8-Diazafluoren-9-one (DFO) is used in forensic science to detect latent fingerprints on paper and other materials. DFO forms highly fluorescent derivatives with amino acids (excitation ~470 nm; emission ~570 nm).

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Reagent forming highly fluorescent derivatives with amino acids (excitation ~470 nm; emission ~570 nm). Used for the detection of latent fingerprints on paper with high sensitivity.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aneta Lewkowicz et al.
Materials (Basel, Switzerland), 13(13) (2020-07-10)
The investigation of innovative label-free α-amino acids detection methods represents a crucial step for the early diagnosis of several diseases. While 1,8-diazafluoren-9-one (DFO) is known in forensic application because of the fluorescent products by reacting with the amino acids present
C.A. Pounds et al.
Journal of Forensic Sciences, 35, 169-169 (1990)
D Wilkinson
Forensic science international, 109(2), 87-103 (2000-03-08)
This paper describes the study of the reaction between 1, 8-diazafluoren-9-one (DFO) with the amino acid L-alanine in methyl alcohol. Particular interest was paid to the possible role of the solvent which appears to react with the DFO to form
Danna E Bicknell et al.
Journal of forensic sciences, 53(5), 1108-1116 (2008-07-22)
1,2-Indanedione belongs to a class of compounds which have demonstrated great potential in the processing of latent prints, particularly in the area of fluorescence. However, variability in results achieved worldwide has precluded it from being used extensively. In order to
Kelly Mayse et al.
Science & justice : journal of the Forensic Science Society, 59(3), 349-358 (2019-05-06)
A study into the modification of 1,8-diazafluoren-9-one (DFO) formulations by the additions of metal salts into the working solution is reported. Similar additions have been found to increase the fluorescence of marks developed using other amino acid reagents including 1,2-indandione

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service