Skip to Content
Merck
All Photos(1)

Key Documents

30434

Sigma-Aldrich

Dansylhydrazine

BioReagent, suitable for fluorescence, ≥90% (HPLC)

Synonym(s):

5-(Dimethylamino)-1-naphthalenesulfonic hydrazide, 5-(Dimethylamino)naphthalene-1-sulfono­hydra­zide, Dns-Hz

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2NC10H6SO2NHNH2
CAS Number:
Molecular Weight:
265.33
Beilstein:
2868662
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.32

product line

BioReagent

Assay

≥90% (HPLC)

mp

126-130 °C (lit.)

solubility

ethanol: soluble

fluorescence

λex 340 nm; λem 525 nm in ethanol

suitability

suitable for fluorescence

storage temp.

2-8°C

SMILES string

CN(C)c1cccc2c(cccc12)S(=O)(=O)NN

InChI

1S/C12H15N3O2S/c1-15(2)11-7-3-6-10-9(11)5-4-8-12(10)18(16,17)14-13/h3-8,14H,13H2,1-2H3

InChI key

KPQYDVAFRDWIBW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Dansylhydrazine (DH) is widely used as a fluorometric reagent for carbonyl compounds. Dansylated carbonyl molecules can be separated and identified by analytical techniques.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Fluorescent marker for carbonyl compounds

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Cheuk Fai Chiu et al.
The journal of physical chemistry. A, 115(34), 9579-9584 (2011-07-20)
The development of chromophore-carbon nanotube hybrids requires efficient and accurate methods to investigate their photophysical properties. Using the ability of the fluorescence labeling of surface species (FLOSS) technique to determine the density of covalently attached dyes to the surface of
Heather L Lord et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 877(13), 1292-1298 (2009-01-07)
Analytical derivatization (AD) increases the sensitivity of analysis by one to three orders of magnitude, stabilizes labile analytes and converts them into readily extractable products. Using a variant of this technique, we applied solid phase analytical derivatization (SPAD) to fully
Osama Y Al-Dirbashi et al.
Biomedical chromatography : BMC, 22(11), 1181-1185 (2008-07-25)
We describe an improved diagnostic method for tyrosinemia type 1 based on quantifying succinylacetone in dried blood spots by ultra-performance liquid chromatography tandem mass spectrometry. Succinylacetone extracted from a single 3/16 inch disk of specimen collection paper containing a dried
1-Dimethylaminonaphthalene-5-sulfonyl hydrazine (dansyl hydrazine): a fluorometric reagent for carbonyl compounds.
R Chayes et al.
Analytical biochemistry, 42(1), 283-286 (1971-07-01)
J M Anderson
Analytical biochemistry, 152(1), 146-153 (1986-01-01)
Methods for the determination of carbonyl compounds of biological origin by high-performance liquid chromatography were improved by the use of new fluorescent derivatizing agents. Eight fluorescent hydrazides were either synthesized or obtained commercially and compared to dansyl hydrazine (1-dimethylaminonaphthalene-5-sulfonylohydrazide). Four

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service