Skip to Content
Merck
All Photos(1)

Key Documents

30030

Sigma-Aldrich

Cymarin

≥96% (HPLC)

Synonym(s):

K-Strophanthidin-D-cymaroside, K-Strophanthin-α

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H44O9
CAS Number:
Molecular Weight:
548.66
Beilstein:
101370
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥96% (HPLC)

SMILES string

CO[C@H]1C[C@@H](O[C@H](C)[C@H]1O)O[C@H]2CC[C@]3(C=O)C4CC[C@]5(C)[C@H](CC[C@]5(O)C4CC[C@]3(O)C2)C6=CC(=O)OC6

InChI

1S/C30H44O9/c1-17-26(33)23(36-3)13-25(38-17)39-19-4-9-28(16-31)21-5-8-27(2)20(18-12-24(32)37-15-18)7-11-30(27,35)22(21)6-10-29(28,34)14-19/h12,16-17,19-23,25-26,33-35H,4-11,13-15H2,1-3H3/t17-,19+,20-,21+,22-,23+,25+,26-,27-,28+,29+,30+/m1/s1

InChI key

XQCGNURMLWFQJR-ZNDDOCHDSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Cymarin was used to study the effect of cardiac glycosides on vascular smooth muscles and ion transport.

Biochem/physiol Actions

Cymarin is a cardiac glycoside and is classified as cardiotonic steroid. It inhibits ion transport by decreasing the activity of (Na-K)-ATPase.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Z B Skorzova et al.
Sudebno-meditsinskaia ekspertiza, 32(2), 35-36 (1989-04-01)
Private technique of extraction isolation and purification, chromatographic detection and photometric determination of zimarin in urine is suggested. Detection limit is 0.01 mg, determination limit is 0.1 mg of glycoside in 100 ml of urine. Method makes it possible to
E V Lopatina et al.
Bulletin of experimental biology and medicine, 146(6), 744-746 (2009-06-11)
We studied the role of cardiac glycosides in the regulation of the growth of retinal tissue explants from 10-12-day-old chicken embryos in organotypic culture. The studied compounds produced a dose-dependent effect on cell proliferation in retinal tissue explants. Ouabain (10(-13)M)
Iu I Pivovarov et al.
Biulleten' eksperimental'noi biologii i meditsiny, 96(8), 24-28 (1983-08-01)
Experiments on rats were made to study the character of rhythmical activity of intact heart and heart with abnormal reactivity under electrical stimulation of the blue spot (BS) and formation in it of the generator of pathologically enhanced excitation (GPEE)
C Giunta et al.
General pharmacology, 16(3), 183-188 (1985-01-01)
Ouabain and K-strophanthoside promote an enhancement of Na+/K+-ATPase activity in a range of cardioglycoside concentrations from 100 nM to 100 pM, with a maximum (+30%) between 10 and 4 nM. Binding experiments with [3H]ouabain show upward-curved Scatchard plots and evidence
Increased in vivo synthesis of retinal Na pump isoforms after pre-treatment with citrate buffer.
S C Specht et al.
Progress in clinical and biological research, 268B, 143-147 (1988-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service