Skip to Content
Merck
All Photos(1)

Key Documents

247M-9

Sigma-Aldrich

EMA (E29) Mouse Monoclonal Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

100
500

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

E29, monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (247M-94)
vial of 0.5 mL concentrate (247M-95)
bottle of 1.0 mL predilute (247M-97)
vial of 1.0 mL concentrate (247M-96)
bottle of 7.0 mL predilute (247M-98)

manufacturer/tradename

Cell Marque®

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100-1:500

isotype

IgG2aκ

control

breast

shipped in

wet ice

storage temp.

2-8°C

visualization

cytoplasmic, membranous

Gene Information

human ... MUC1(4582)

General description

Anti-EMA antibody is a useful marker for staining many carcinomas. It stains normal and neoplastic cells from various tissues, including mammary epithelium, sweat glands and squamous epithelium. Hepatocellular carcinoma, adrenal carcinoma and embryonal carcinomas are consistently EMA negative, so keratin positivity with negative EMA favors one of these tumors. EMA is frequently positive in meningioma, which can be useful when distinguishing it from other intracranial neoplasms, e.g. Schwannomas. The absence of EMA can also be of value since negative EMA staining is characteristic of some tumors including adrenal carcinoma, seminomas, paraganglioma and hepatoma.

Quality


IVD

IVD

IVD

RUO

Linkage

EMA Positive Control Slides, Product No. 247S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: [email protected]

Legal Information

Cell Marque is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W H Redding et al.
Lancet (London, England), 2(8362), 1271-1274 (1983-12-03)
An immunocytochemical method was used to screen smears obtained at primary surgery from multiple bone-marrow sites in 110 patients with breast cancer; at this time other techniques did not reveal metastases. Tumour cells were detected in the bone-marrow of 31
T W Beer et al.
Histopathology, 37(3), 218-223 (2000-09-06)
Seventy-five skin tumours were studied to investigate the value of immunohistochemistry in differentiating basal cell, squamous cell and basosquamous carcinomas of the skin. Archived paraffin-embedded tissue samples of basal cell carcinomas (n = 39), squamous cell carcinomas (n = 23)
D P Dearnaley et al.
British journal of cancer, 44(1), 85-90 (1981-07-01)
We have developed a technique for the immunocytochemical staining of marrow smears using antiserum to epithelial membrane antigen (EMA). This membrane component is confined to, but widely distributed in, epithelial tissues and tumours derived from them, and is strongly expressed
Adam Wahida et al.
iScience, 26(10), 107879-107879 (2023-10-23)
Renal ischemia-reperfusion injury (IRI) is associated with reduced allograft survival, and each additional hour of cold ischemia time increases the risk of graft failure and mortality following renal transplantation. Receptor-interacting protein kinase 3 (RIPK3) is a key effector of necroptosis
M Fraga et al.
American journal of clinical pathology, 103(1), 82-89 (1995-01-01)
Bone marrow involvement by anaplastic large cell anaplastic large cell (ALC) lymphoma can be difficult to detect on routine morphologic examination alone. In a series of 42 patients with ALC lymphoma, the authors analyzed: (1) the usefulness of a limited

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service