24340C
EX-CELL® 325 PF CHO Serum-Free Medium for CHO Cells
protein free, dry powder, suitable for cell culture
Pharma Manufacturing
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
description
for research or for further manufacturing use
without L-glutamine
Quality Level
form
dry powder
technique(s)
cell culture | mammalian: suitable
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
General description
EX-CELL® 325 PF CHO is a protein-free medium which has been developed for the growth of Chinese Hamster Ovary (CHO) cells. Because it contains no large macromolecules, EX-CELL 325 PF CHO facilitates the isolation and purification of secreted proteins from the cells. CHO cells propagated in EX-CELL 302 serum-free media can be transferred directly into this protein-free medium without extensive weaning protocols. This medium is supplied without L-glutamine and does not contain purines or pyrimidines to provide an appropriate medium for specialized CHO cell lines (i.e., Glutamine Synthetase, or the GS System™ and DHFR- selection systems).
Quantity
Formulated to contain 21.0 grams of powder per liter of medium.
Reconstitution
Aseptically add 20-40 ml of 200 mM L-glutamine solution per liter of medium prior to use.
Legal Information
EX-CELL is a registered trademark of Merck KGaA, Darmstadt, Germany
GS System is a trademark of Lonza Group Ltd.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service