Skip to Content
Merck
All Photos(1)

Key Documents

21742

Sigma-Aldrich

4-Nitrophenyl octanoate

≥90.0% (GC), liquid

Synonym(s):

4-Nitrophenyl caprylate, Caprylic acid 4-nitrophenyl ester, Octanoic acid 4-nitrophenyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H19NO4
CAS Number:
Molecular Weight:
265.30
Beilstein:
8989916
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.83

product name

4-Nitrophenyl octanoate, ≥90.0% (GC)

Quality Level

Assay

≥90.0% (GC)

form

liquid

refractive index

n20/D 1.510

density

1.095 g/mL at 20 °C (lit.)

storage temp.

2-8°C

SMILES string

CCCCCCCC(=O)Oc1ccc(cc1)[N+]([O-])=O

InChI

1S/C14H19NO4/c1-2-3-4-5-6-7-14(16)19-13-10-8-12(9-11-13)15(17)18/h8-11H,2-7H2,1H3

InChI key

GGIDEJQGAZSTES-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Yakai Lin et al.
Membranes, 10(8) (2020-07-29)
The rational design and realization of multiscale porous structures has been a long-standing challenge in membrane science. Block copolymers (BCPs) with their self-assembly-enabled nanodomains have the potential to make structural breakthroughs. An amphipathic Janus membrane, with a hierarchical multiscale hyperporous
Priyatharini Ambigaipalan et al.
Food chemistry, 221, 1883-1894 (2016-12-17)
Phenolics from free and hydrolyzed fractions of pomegranate juice (PJ) and seeds (PS) were evaluated. In general, total phenolic contents and scavenging of ABTS+, DPPH and hydroxyl radicals, as well as metal chelation of the soluble fraction from PS, were
Thorsten Eggert et al.
FEMS microbiology letters, 225(2), 319-324 (2003-09-03)
Bacillus subtilis secretes two lipases LipA and LipB into the culture medium. Both enzyme genes were differentially expressed depending on the growth conditions as determined by activity assays and Western blotting in B. subtilis mutant strains lipA, lipB, and the
Simona Fabroni et al.
Journal of the science of food and agriculture, 96(14), 4713-4723 (2016-03-13)
The phytotherapic treatment of overweight and/or moderate obesity is growing widely, thus there is a great interest towards the phenolic compounds of fruits and vegetables which may inhibit pancreatic lipase enzyme. In this study, we report the chemical composition and
Mariangela Marrelli et al.
Plants (Basel, Switzerland), 9(1) (2020-01-16)
Lobularia maritima (L.) Desv. is a perennial herb growing wild in the Mediterranean basin. The aim of this work was to assess the fatty acid, terpene, phytosterol, and phenolic composition of the methanolic extract and its sub-fractions using Gas Chromatography-Mass

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service