213-500
Endothelial Cell Growth Medium v2 (500 ml)
Synonym(s):
Endothelial Cell Culture Medium
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352207
NACRES:
NA.81
Recommended Products
form
liquid
shelf life
6 mo.
manufacturer/tradename
Cell Applications, Inc
shipped in
wet ice
storage temp.
2-8°C
General description
Endothelial Cell Growth Medium v2 (500 ml) is an all-in-one-ready-to-use media that contains growth factors that facilitate accelerated growth.
Application
Endothelial Cell Growth Medium v2 (500 ml) has been used to culture:
- human aortic endothelial cells
- human cardiac microvascular endothelial cells
- human coronary artery endothelial cells
Quality
Each lot was tested for the ability to support plating, spreading and proliferation
Preparation Note
contained in protocol
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service