Skip to Content
Merck
All Photos(1)

Key Documents

21210

Sigma-Aldrich

D-Pantothenic acid hemicalcium salt

≥98.0%

Synonym(s):

(R)-(+)-N-(2,4-Dihydroxy-3,3-dimethyl-1-oxobutyl)-β-alanine hemicalcium salt, Calcium D-pantothenate, Vitamin B5

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2C(CH3)2CH(OH)CONHCH2CH2CO2 ·1/2Ca
CAS Number:
Molecular Weight:
238.27
Beilstein:
3769272
EC Number:
MDL number:
UNSPSC Code:
12352106
eCl@ss:
34058012
PubChem Substance ID:
NACRES:
NA.79

biological source

synthetic

Quality Level

Assay

≥98.0%

form

powder or crystals

optical activity

[α]20/D +27±2°, c = 5% in H2O

impurities

≤0.002% heavy metals

loss

≤3% loss on drying

color

white

pH

6.8-7.2 (25 °C, 50 mg/mL in H2O)

solubility

H2O: 50 mg/mL at 25 °C, clear, almost colorless

SMILES string

CC(C)(CO)[C@@H](O)C(=O)NCCC(=O)O[Ca]OC(=O)CCNC(=O)[C@H](O)C(C)(C)CO

InChI

1S/2C9H17NO5.Ca/c2*1-9(2,5-11)7(14)8(15)10-4-3-6(12)13;/h2*7,11,14H,3-5H2,1-2H3,(H,10,15)(H,12,13);/q;;+2/p-2/t2*7-;/m00./s1

InChI key

FAPWYRCQGJNNSJ-UBKPKTQASA-L

Looking for similar products? Visit Product Comparison Guide

General description

D-Pantothenic acid hemicalcium salt, also referred as calcium D-pantothenate, is a water soluble B complex vitamin. It is present in a variety of foods.

Application

D-Pantothenic acid hemicalcium salt has been used as a supplement in complete media to culture Aspergillus-nidulans.
Precursor in the biosynthesis of coenzyme A.

Biochem/physiol Actions

D-Panthenol is the alcohol analog and biological precursor of D-pantothenic acid. These analogs are precursors in the biosynthesis of the phosphopantetheine moiety of coenzyme A. D-Panthenol and D-Pantothenic acid are used in a variety of skin protection products. D-pantothenic acid may have a role in controlling keratinocyte proliferation and differentiation. D-Panthenol may be used for studies of skin protection emulsions from UV irradiation and as a humectant. D-pantothenic acid is crucial for normal epithelial function. It facilitates wound healing process.

Physical properties

Due to the unstable, hygroscopic nature of the free acid, the calcium salt is employed.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Pantothenic acid
Kelly GS
Alternative Medicine Review, 16(3), 263-275 (2011)
Transport Assays in Aspergillus nidulans
Krypotou E and Diallinas G
Molecular Microbiology (2013)
Pantothenic acid
Rawalpally TR
Kirk-Othmer Encyclopedia of Chemical Technology (2000)
Organisation of the pantothenate (vitamin B5) biosynthesis pathway in higher plants
Ottenhof HH, et al.
The Plant Journal, 37(1), 61-72 (2004)
Wound healing activity of Memecylon umbellatum Burm
Puratchikody A and Nagalakshmi G
Journal of plant sciences, 2(2), 179-186 (2007)

Articles

Enzyme Reagent Coenzyme A (CoA, CoASH or HSCoA) is the key cofactor in first step of the TCA cycle, responsible for transferring the acetyl group from pyruvate oxidation to oxaloacetate yielding citrate. Available through Sigma-Aldrich online.

Enzyme Reagent Coenzyme A (CoA, CoASH or HSCoA) is the key cofactor in first step of the TCA cycle, responsible for transferring the acetyl group from pyruvate oxidation to oxaloacetate yielding citrate. Available through Sigma-Aldrich online.

Enzyme Reagent Coenzyme A (CoA, CoASH or HSCoA) is the key cofactor in first step of the TCA cycle, responsible for transferring the acetyl group from pyruvate oxidation to oxaloacetate yielding citrate. Available through Sigma-Aldrich online.

Enzyme Reagent Coenzyme A (CoA, CoASH or HSCoA) is the key cofactor in first step of the TCA cycle, responsible for transferring the acetyl group from pyruvate oxidation to oxaloacetate yielding citrate. Available through Sigma-Aldrich online.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service