Skip to Content
Merck
All Photos(1)

Key Documents

17927

Sigma-Aldrich

9-Oxo-10(9H)-acridineacetic acid

for HPLC derivatization, ≥99.0% (T)

Synonym(s):

(9,10-Dihydro-9-oxo-10-acridinyl)acetic acid, 10-(Carboxymethyl)-9(10H)acridone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H11NO3
CAS Number:
Molecular Weight:
253.25
Beilstein:
227508
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.32

grade

for HPLC derivatization

Quality Level

Assay

≥99.0% (T)

form

solid

mp

~289 °C (dec.)

SMILES string

OC(=O)CN1c2ccccc2C(=O)c3ccccc13

InChI

1S/C15H11NO3/c17-14(18)9-16-12-7-3-1-5-10(12)15(19)11-6-2-4-8-13(11)16/h1-8H,9H2,(H,17,18)

InChI key

UOMKBIIXHQIERR-UHFFFAOYSA-N

Application

9-Oxo-10(9H)-acridineacetic acid and its analogs are used to study their potential activity as interferon inducers.

Other Notes

Reagent for pre-column derivatization of amino acids for fluorescent determination in HPLC; Building block in pharmaceutics

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

V M Frolov et al.
Likars'ka sprava, (1-2)(1-2), 102-107 (2012-10-06)
The effect of the immunotropic drug cycloferon and herbal medicine resources on the basis of Cynara scolimus L. on the blood cytokine profile in the patients with chronic viral hepatitis C (CVHC) in medical rehabilitation (MR) period. Established that prior
S.A. Lyakhov et al.
Pharmazie, 49, 367-367 (1994)
A V Gordeets et al.
Eksperimental'naia i klinicheskaia farmakologiia, 76(2), 17-19 (2013-05-02)
The case records of 120 children older than 4 years with rotavirus infection have been studied. In this group, 22 patients received basic therapy and 98 received complex therapy with cycloferon. Appropriate therapy resulted in rapid positive dynamics of clinical
E D Bazhanova
Eksperimental'naia i klinicheskaia farmakologiia, 75(10), 42-46 (2012-12-18)
A detailed analysis of the literature data gives contradictory information about the role of interferon-alpha in the regulation of apoptosis, while there are almost no data on the participation of cycloferon in this process. Results of original experiments in recent
V M Frolov et al.
Antibiotiki i khimioterapiia = Antibiotics and chemoterapy [sic], 57(5-6), 18-27 (2012-11-20)
The use of reamberin and cycloferon combination for increasing the antibacterial therapy efficacy in patients with chronic sepsis (CS) was studied. It was shown that the combination provided more rapid elimination of the CS exacerbation symptoms, normalization of the hematologic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service