Skip to Content
Merck
All Photos(1)

Key Documents

16001

Sigma-Aldrich

Brij® L4

main component: tetraethylene glycol dodecyl ether

Synonym(s):

Polyethylene glycol dodecyl ether

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Beilstein:
1900171
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:

description

non-ionic

technique(s)

protein quantification: suitable

refractive index

n20/D 1.453

HLB value

9.7

density

~0.95 g/mL at 20 °C

SMILES string

CCCCCCCCCCCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCO

InChI

1S/C58H118O24/c1-2-3-4-5-6-7-8-9-10-11-13-60-15-17-62-19-21-64-23-25-66-27-29-68-31-33-70-35-37-72-39-41-74-43-45-76-47-49-78-51-53-80-55-57-82-58-56-81-54-52-79-50-48-77-46-44-75-42-40-73-38-36-71-34-32-69-30-28-67-26-24-65-22-20-63-18-16-61-14-12-59/h59H,2-58H2,1H3

InChI key

IEQAICDLOKRSRL-UHFFFAOYSA-N

Caution

product may precipitate on storage, resolve at 50°C

Legal Information

Brij is a registered trademark of Croda International PLC

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Erkin et al.
European journal of vascular and endovascular surgery : the official journal of the European Society for Vascular Surgery, 44(6), 593-597 (2012-11-06)
Foam sclerosants are widely used in sclerotherapy and have been accepted as more effective than the liquid form; however, there is no consensus about the most applicable and effective concentration. The aim of this study was to investigate the histopathological
Otte Brosjö et al.
Acta orthopaedica, 84(5), 502-505 (2013-11-01)
Recent data suggest that percutaneous sclerotherapy is a safe alternative to surgery for treatment of aneurysmal bone cysts (ABCs). We present our experience of this method. We retrospectively analyzed data from 38 consecutive patients treated with repeated injections of polidocanol.
Jun Koizumi et al.
AJR. American journal of roentgenology, 199(1), 200-207 (2012-06-27)
Balloon-occluded retrograde transvenous obliteration has been traditionally based on liquid sclerotherapy. However, overdose and systemic spillage of liquid sclerosant can cause severe complications, such as hemolysis, which lead to hemoglobinuria, allergy, acute respiratory distress syndrome, and other disorders. The purpose
Darío Morais et al.
Rhinology, 50(1), 80-86 (2012-04-04)
Hereditary Haemorrhagic Telangiectasia or Rendu-Osler-Weber syndrome is a rare autosomal dominant vascular disease characterized by mucocutaneous and gastrointestinal telangiectases and localized arteriovenous malformations in lung, brain and liver. Epistaxis, due to rupture of telangiectases of the nasal mucosa, is the
Jacomina W van den Esschert et al.
Annals of surgery, 255(2), 311-318 (2012-01-14)
To compare the safety and hypertrophy response after portal vein embolization (PVE) using 2 absorbable and 3 permanent embolization materials. Portal vein embolization is used to increase future remnant liver volume preoperatively. Application of temporary, absorbable embolization materials could be

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service