Skip to Content
Merck
All Photos(1)

Key Documents

15326

Sigma-Aldrich

Lead(II) citrate tribasic trihydrate

purum, suitable for electron microscopy

Synonym(s):

Citric acid lead salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5O7)2Pb3 · 3H2O
CAS Number:
Molecular Weight:
1053.85
Beilstein:
3920957
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.25

grade

purum

Quality Level

form

powder

technique(s)

electron microscopy: suitable

SMILES string

O.O.O.OC2(CC(=O)O[PbH2]OC(=O)CC1(O)CC(=O)O[PbH2]OC1=O)CC(=O)O[PbH2]OC2=O

InChI

1S/2C6H8O7.3H2O.3Pb/c2*7-3(8)1-6(13,5(11)12)2-4(9)10;;;;;;/h2*13H,1-2H2,(H,7,8)(H,9,10)(H,11,12);3*1H2;;;/q;;;;;3*+2/p-6

InChI key

GMPMGSCJCDAUMP-UHFFFAOYSA-H

Looking for similar products? Visit Product Comparison Guide

Application

Citric acid lead salt has been used in a study to assess relationships between erosive damage to teeth and high intake of sports drinks. Citric acid lead salt has also been used in a study to investigate the effects of low-molecular-weight organic acids (LMWOAs) and residence time on desorption of Cu, Cd, and Pb from two typical Chinese soils.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 1A - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Novel biodegradable cholesterol-modified polyrotaxane hydrogels for cartilage regeneration
Tachaboonyakiat, W., et al.
Chemosphere, 15, 1389-1404 (2004)
Effects of low-molecular-weight organic acids and residence time on desorption of Cu, Cd, and Pb from soils
Qin, F., et al.
Journal of the California Dental Association, 57, 253-263 (2004)
Sports drinks and dental erosion
Noble, W., et al.
Journal of Biomaterials Science. Polymer Edition, 39, 233-238 (2011)
Zu-Hong He et al.
Redox biology, 28, 101364-101364 (2019-11-16)
Inflammation is a self-defense response to protect individuals from infection and tissue damage, but excessive or persistent inflammation can have adverse effects on cell survival. Many individuals become especially susceptible to chronic-inflammation-induced sensorineural hearing loss as they age, but the
Hang Yu Wang et al.
Journal of cellular physiology, 235(3), 2722-2737 (2019-09-12)
Liver fibrosis (LF) is the result of a vicious cycle between inflammation-induced chronic hepatocyte injury and persistent activation of hepatic stellate cells (HSCs). Mesenchymal stem cell (MSC)-based therapy may represent a potential remedy for treatment of LF. However, the fate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service