Skip to Content
Merck
All Photos(1)

Key Documents

14853

Sigma-Aldrich

BES

BioXtra, for molecular biology, ≥99.5% (T)

Synonym(s):

N,N-Bis(2-hydroxyethyl)-2-aminoethanesulfonic acid, N,N-Bis(2-hydroxyethyl)taurine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H15NO5S
CAS Number:
Molecular Weight:
213.25
Beilstein:
1781572
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:

grade

for molecular biology

product line

BioXtra

Assay

≥99.5% (T)

form

powder or crystals

impurities

DNases, none detected
RNases, none detected
insoluble matter, passes filter test
phosphatases, none detected
proteases, none detected

ign. residue (900 °C)

≤0.05%

pH

2.5-5.0 (25 °C, 1 M in H2O)

useful pH range

6.4-7.8

pKa (25 °C)

7.1

solubility

H2O: 1 M at 20 °C, clear, colorless

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤100 mg/kg

cation traces

Al: ≤5 mg/kg
As: ≤0.1 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

λ

1 M in H2O

UV absorption

λ: 260 nm Amax: 0.095
λ: 280 nm Amax: 0.080

SMILES string

OCCN(CCO)CCS(O)(=O)=O

InChI

1S/C6H15NO5S/c8-4-1-7(2-5-9)3-6-13(10,11)12/h8-9H,1-6H2,(H,10,11,12)

InChI key

AJTVSSFTXWNIRG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

BES, or N,N-Bis(2-hydroxyethyl)-2-aminoethanesulfonic acid, can be used to study molecular biology, biological buffers, molecular biology reagents and zwitterionic compounds. BES has been used to study versatile catalyst precursors for mild hydrocarboxylation of alkanes to carboxylic acids. BES has also been used to determine that saline and buffers minimize the action of interfering factors in the bacterial endotoxins test.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marina V Kirillova et al.
Inorganic chemistry, 51(9), 5224-5234 (2012-04-10)
The facile aqueous medium reactions of copper(II) nitrate with BES biobuffer [(HOCH(2)CH(2))(2)N(CH(2)CH(2)SO(3)H), hereinafter referred as H(3)bes] in the presence of various benzenecarboxylic acids [benzoic (Hba), 3-hydroxybenzoic (Hhba), and 3,5-dihydroxybenzoic (Hdhba) acid] and lithium hydroxide gave rise to the self-assembly generation
Kensuke Nishimiya et al.
Circulation journal : official journal of the Japanese Circulation Society, 79(6), 1323-1331 (2015-04-07)
Coronary adventitia harbors a wide variety of components, such as inflammatory cells and vasa vasorum (VV). Adventitial VV initiates the development of coronary artery diseases as an outside-in supply route of inflammation. We have recently demonstrated that drug-eluting stent implantation
Russell A Jensen et al.
The journal of physical chemistry letters, 6(15), 2933-2937 (2015-08-13)
Colloidal CdSe quantum dot (QD) core ensembles were photodimmed and allowed to recover in the dark using ambient thermal energy at a range of temperatures. Nonlinear thermal recovery is well described by a stretched exponential function, and further analysis yields
Yingjie Zhang et al.
Nano letters, 15(7), 4657-4663 (2015-06-24)
Noncrystalline semiconductor materials often exhibit hysteresis in charge transport measurements whose mechanism is largely unknown. Here we study the dynamics of charge injection and transport in PbS quantum dot (QD) monolayers in a field effect transistor (FET). Using Kelvin probe
C A Poole et al.
In vitro, 18(9), 755-765 (1982-09-01)
Chick embryo epiphyseal chondrocytes cultured in media containing HEPES, TES, and BES zwitterion buffers, used in combination or independently, consistently developed cytoplasmic vacuoles. This cytoplasmic vacuolation was resolved when the zwitterion buffered media was replaced by media containing bicarbonate:CO2 enriched

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service