Skip to Content
Merck
All Photos(1)

Key Documents

14470

Sigma-Aldrich

1,4-Bis(acryloyl)piperazine

BioXtra, suitable for electrophoresis, ≥99.0% (TLC)

Synonym(s):

1,4-Di(acryloyl)piperazine, N,N′-Bis(acryloyl)piperazine, BAP, PDA, Piperazine diacrylamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14N2O2
CAS Number:
Molecular Weight:
194.23
Beilstein:
745431
MDL number:
UNSPSC Code:
41105319
PubChem Substance ID:
NACRES:
NA.26

product line

BioXtra

Quality Level

Assay

≥99.0% (TLC)

form

solid

technique(s)

electrophoresis: suitable

impurities

≤0.05% free acrylic acid

mp

91.5-93.5 °C (lit.)

anion traces

chloride (Cl-): ≤200 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

suitability

suitable for electrophoresis

storage temp.

2-8°C

SMILES string

C=CC(=O)N1CCN(CC1)C(=O)C=C

InChI

1S/C10H14N2O2/c1-3-9(13)11-5-7-12(8-6-11)10(14)4-2/h3-4H,1-2,5-8H2

InChI key

YERHJBPPDGHCRJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Fabrication and application of zwitterionic phosphorylcholine functionalized monoliths with different hydrophilic crosslinkers in hydrophilic interaction chromatography: This study demonstrates the effective use of 1,4-Bis(acryloyl)piperazine as a crosslinker in creating zwitterionic monoliths for chromatographic applications, highlighting its utility in enhancing polymer interactions and selectivity in separation processes (Wang et al., 2020).

  • Poly(amidoamine) conjugates with disulfide-linked cholesterol pendants self-assembling into redox-sensitive nanoparticles: This application uses a derivative concept where 1,4-Bis(acryloyl)piperazine could theoretically enhance the self-assembly and stability of redox-sensitive polymeric nanoparticles, offering insights into controlled release and targeted drug delivery systems (Ranucci et al., 2008).

Other Notes

Used as a vinylating agent for graft-copolymerization of different enzymes to cellulose, dextran, or starch; New crosslinking agent for polyacrylamide gels with improved protein separation and detection by silver staining; Increased resolution in 2D-protein electrophoresis.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Adam C McDonnell et al.
Experimental physiology, 104(8), 1250-1261 (2019-07-06)
What is the central question of this study? It is well established that muscle and bone atrophy in conditions of inactivity or unloading, but there is little information regarding the effect of a hypoxic environment on the time course of
Sayaka Nose-Ogura et al.
Scandinavian journal of medicine & science in sports, 30(8), 1379-1386 (2020-04-15)
The effects of transdermal estradiol treatment (HT) in amenorrheic athletes (AA) with low body weight (BW) and low bone mineral density (BMD) are unknown. To investigate whether HT increases BMD in AA with low BW and to compare the results
L. D'Angiuro et al.
Angew. Makromol. Chem., 104, 129-129 (1982)
K R Hughes et al.
Journal of speech, language, and hearing research : JSLHR, 62(7), 2386-2399 (2019-06-30)
Purpose Infant siblings of children with autism spectrum disorder (ASIBs) and infants with fragile X syndrome (FXS) are both at risk for developing autism spectrum disorder (ASD) and communication disorders; however, very few studies have examined 1 of the earliest
Chun-Min Kang et al.
Journal of microbiology, immunology, and infection = Wei mian yu gan ran za zhi, 53(6), 882-891 (2020-04-20)
Rapid and accurate identification of pathogens and their antibiotic resistance directly from flagged blood cultures can aid clinicians in optimizing early antibiotic treatment and improve the clinical outcomes, especially in settings associated with high rates of bloodstream infection caused by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service