Skip to Content
Merck
All Photos(4)

Key Documents

P1767

Sigma-Aldrich

Potassium hydroxide

≥85% KOH basis, pellets, white

Synonym(s):

Caustic potash

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KOH
CAS Number:
Molecular Weight:
56.11
EC Number:
MDL number:
UNSPSC Code:
12352106
eCl@ss:
38100303
PubChem Substance ID:
NACRES:
NA.21
Assay:
≥85% KOH basis
form:
pellets

vapor pressure

1 mmHg ( 719 °C)

Quality Level

Assay

≥85% KOH basis

form

pellets

impurities

≤2.0% K2CO3

color

white

pH

~13.5 (25 °C, 5.6 g/L)

mp

361 °C (lit.)

SMILES string

[OH-].[K+]

InChI

1S/K.H2O/h;1H2/q+1;/p-1

InChI key

KWYUFKZDYYNOTN-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Potassium hydroxide, also known as caustic potash, is a water soluble strong base. The electron scattering in KOH has been investigated. Its infrared spectrum has been recorded and analyzed. The impact of surface functionalization of active carbons using KOH has been studied. Its role as a catalyst in the Michael addition reaction has been explored.

Application

Potassium hydroxide (KOH) has been used to prepare methanolic-KOH that is employed in a protocol for saponification of fruit extracts.
KOH has been used for adjusting pH during the preparation of the following:
  • Murashige and Skoog (MS) agar plates
  • MS liquid medium
  • MES ((2-(N-morpholino)-ethanesulfonic acid hydrate) solution
  • microtubule stabilizing buffer (MTSB)
  • 4% paraformaldehyde fixative (PFA)

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vibrational spectrum of crystalline potassium hydroxide.
Snyder RG, et al.
J. Chem. Phys. , 33(4), 1171-1177 (1960)
Carotenoid Extraction and Quantification from Capsicum annuum.
Richins RD, et al.
Bio-protocol, e1256-e1256 (2014)
Michael addition catalyzed by potassium hydroxide under ultrasound.
Li JT, et al.
Synthetic Communications, 33(3), 353-359 (2003)
Electron impact study of potassium hydroxide.
Vuskovic L and Trajmar S.
J. Chem. Phys. , 71(9), 3887-3889 (1979)
Potassium hydroxide modified active carbon for adsorptive refrigerators.
Buczek B and Wolak E.
Adsorption, 14(2-3), 283-287 (2008)

Protocols

This procedure may be used for all Catalase products.

This procedure may be used for all Ficin products.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service