Skip to Content
Merck
All Photos(2)

Key Documents

C3136

Sigma-Aldrich

Cesium sulfate

Grade I, ≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Cs2O4S
CAS Number:
Molecular Weight:
361.87
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.55
grade:
for analytical purposes
form:
powder

grade

for analytical purposes

type

Grade I

Assay

≥99%

form

powder

pH

6-7.5 (25 °C, 723.7 g/L)

mp

1019 °C (lit.)

density

4.243 g/mL at 25 °C (lit.)

anion traces

sulfate (SO42-): 26.1-26.9%

absorption

≤0.2 at 280 in H2O at 50%

SMILES string

[Cs+].[Cs+].[O-]S([O-])(=O)=O

InChI

1S/2Cs.H2O4S/c;;1-5(2,3)4/h;;(H2,1,2,3,4)/q2*+1;/p-2

InChI key

FLJPGEWQYJVDPF-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application



  • Catalytic effect of cesium on the oxidation behavior of cation exchange resins in Li(2)CO(3)-Na(2)CO(3)-K(2)CO(3) melt.: This study explores the catalytic effects of cesium in enhancing the oxidation of cation exchange resins within a carbonate melt, potentially applicable in recycling and waste management practices (Zheng et al., 2022).


  • Electron paramagnetic resonance characterization and electron spin relaxation of manganate ion in glassy alkaline LiCl solution and doped into Cs(2)SO(4).: This research provides insights into the properties of manganate ions within cesium sulfate, contributing to the development of new materials for various technological applications (Mahapatro et al., 2022).


  • Response of Arabidopsis halleri to cesium and strontium in hydroponics: Extraction potential and effects on morphology and physiology.: Investigates the phytoremediation potential of Arabidopsis halleri in removing cesium from environments, an important study for environmental cleanup strategies (Burger et al., 2019).


  • Response of Plantago major to cesium and strontium in hydroponics: Absorption and effects on morphology, physiology and photosynthesis.: Discusses how Plantago major plants absorb and respond to cesium, with implications for phytoremediation and environmental management (Burger et al., 2019).


Quality

Yields a clear colorless solution.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Characterization of T4 double-stranded RNA by equilibrium centrifugation in Cs2SO4- and alkaline iodide gradients.
D E Helland
FEBS letters, 117(1), 79-83 (1980-08-11)
Haiyan Liu et al.
Journal of pharmaceutical and biomedical analysis, 42(3), 384-388 (2006-05-30)
In this paper, a simple, rapid and sensitive flow-injection chemiluminescence method has been developed for the determination of metoprolol tartrate, which acts as a kind of sensitizer in the chemiluminescence emission from the redox of SO(3)(2-) with Ce(IV) in acidic
P J Roughley et al.
Biochimica et biophysica acta, 759(1-2), 58-66 (1983-08-23)
Proteoglycan subunits from human articular cartilage were fractionated by caesium sulphate density gradient centrifugation. A single heterogeneous population of molecules was produced whose average density decreased with increasing age of the individual from which they were obtained. At no density
N Isoda et al.
Journal of biochemistry, 90(2), 551-554 (1981-08-01)
Upon CS2SO4 density-gradient equilibrium centrifugation, 60S subunits of rat liver ribosomes were dissociated to form three bands at the densities of 1.55, 1.40, and 1.30 g/ml. The bands at 1.55 and 1.30 g/ml were shown to contain exclusively RNA or
F Bonnet et al.
Biochimica et biophysica acta, 623(1), 57-68 (1980-05-29)
A true isopycnic centrifugation method for the study of the bovine nasal cartilage proteoglycan polydispersity is presented. The use of cesium sulfate as gradient forming salt instead of cesium chloride allowed proteoglycan banding without any sedimentation at the bottom of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service