Skip to Content
Merck
All Photos(5)

Key Documents

A5764

Sigma-Aldrich

Ammonium phosphate dibasic

reagent grade, ≥98.0%

Synonym(s):

Ammonium hydrogenphosphate, Diammonium hydrogenphosphate, di-Ammonium hydrogenphosphate (sec)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NH4)2HPO4
CAS Number:
Molecular Weight:
132.06
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.21

grade

reagent grade

Quality Level

Assay

≥98.0%

form

powder or crystals

pH

7.5-9.0 (25 °C, 132.1 g/L)

mp

155 °C (dec.) (lit.)

solubility

water: soluble 132 g/L at 20 °C

SMILES string

N.N.OP(O)(O)=O

InChI

1S/2H3N.H3O4P/c;;1-5(2,3)4/h2*1H3;(H3,1,2,3,4)

InChI key

MNNHAPBLZZVQHP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Ammonium phosphate dibasic may be used in the following studies:
  • As an activator in generating activated carbon using waste ramulus mori as a source material.
  • To compose the mobile phase for the removal of synthetic impurities from voriconazole by HPLC.
  • Preparation of Eu ions doped oxyapatite Ca8Gd2(PO4)6O2 (CGP) samples.
  • Syringe-type cartridge based electric field-assisted solid phase extraction of sunset yellow, an anionic dye.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation and characterization of activated carbon from waste ramulus mori.
Tang YB, et al.
Chemical Engineering Journal, 203, 19-24 (2012)
Burdock GA.
Encyclopedia of Food and Color Additives, 1, 144-144 (1997)
Study of metastable equilibria in the ternary system NH4H 2PO4-(NH4)2HPO4-H2O at -7 and -9 ?C.
Fluid Phase Equilibria, 354, 47-53 (2013)
Development and validation of a stability-indicating HPLC method for determination of voriconazole and its related substances.
Gu P and Li Y.
Journal of Chromatographic Science, 47(7), 594-598 (2009)
Electric Field-Assisted Solid Phase Extraction: Study of Electrochromatographic Parameters with an Anionic Model Compound.
Orlando RM, et al.
Journal of the Brazilian Chemical Society, 26(2), 310-318 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service