Skip to Content
Merck
All Photos(1)

Key Documents

83352

Sigma-Aldrich

Mercury(II) acetate

puriss. p.a., ACS reagent, ≥99.0% (precipitation titration)

Synonym(s):

Mercuric acetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3COO)2Hg
CAS Number:
Molecular Weight:
318.68
Beilstein:
3563831
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NB.24
Assay:
≥99.0% (precipitation titration)
form:
powder or crystals
solubility:
water: soluble(lit.)

grade

ACS reagent
puriss. p.a.

Quality Level

Assay

≥99.0% (precipitation titration)

form

powder or crystals

ign. residue (after reduction)

≤0.02%

mp

179-182 °C (lit.)

solubility

water: soluble(lit.)

anion traces

chloride (Cl-): ≤50 mg/kg
nitrate (NO3-): ≤50 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤10 mg/kg
Hg+: ≤4000 mg/kg
K: ≤50 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

SMILES string

CC(=O)O[Hg]OC(C)=O

InChI

1S/2C2H4O2.Hg/c2*1-2(3)4;/h2*1H3,(H,3,4);/q;;+2/p-2

InChI key

BRMYZIKAHFEUFJ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Mercury(II) acetate forms various addition compounds with olefins. Polarographic evaluation of these addition compounds has been proposed. It participates in the synthesis of nitrate esters, acetate esters, alcohols and ethers.

Application

Mercury(II) acetate may be used in the preparation of 2-benzylidene-3(2H)-benzofuran-3-ones (aurones).

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Polarography of olefin-mercury (II) acetate addition compounds.
Fleet B and Jee RD.
Journal of Electroanalytical Chemistry, 25(3), 397-408 (1970)
Mercury-assisted solvolyses of alkyl halides: Simple procedures for the preparation of nitrate esters, acetate esters, alcohols and ethers.
McKillop A and Ford ME.
Tetrahedron, 30(15), 2467-2475 (1974)
Synthesis of 2-benzylidene-3 (2H)-benzofuran-3-ones (aurones) by oxidation of 2'-hydroxychalcones with mercury (II) acetate.
Sekizaki H.
Bulletin of the Chemical Society of Japan, 61(4), 1407-1409 (1988)
N Milosevic et al.
European journal of pharmacology, 401(3), 317-328 (2000-08-11)
The role of intercellular signalling between liver cells in lead (Pb)(1)-induced liver toxicity was investigated in cocultures of freshly isolated and cultured rat hepatocytes and Kupffer cells. The Kupffer cells (seeded onto culture dish inserts), the hepatocytes or the two
Yukio Hitotsuyanagi et al.
The Journal of organic chemistry, 67(10), 3266-3271 (2002-05-11)
A novel cis amide bond surrogate incorporating 1,2,4-triazole was designed and synthesized by the reaction of a thionotripeptide, formic hydrazide, and mercury(II) acetate. This method of surrogate formation was also applicable to a cyclic thionopeptide.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service