Skip to Content
Merck
All Photos(1)

Key Documents

77234

Sigma-Aldrich

Perchloric acid

puriss. p.a., ACS reagent, packed in coated, shock- and leak-protected glass bottle, ≥60% (T)

Synonym(s):

PCA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HClO4
CAS Number:
Molecular Weight:
100.46
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.21

grade

ACS reagent
puriss. p.a.

Quality Level

form

liquid

quality

packed in coated, shock- and leak-protected glass bottle

concentration

≥60% (T)

impurities

≤0.00005% free chlorine (Cl)
≤0.001% total nitrogen (N)

ign. residue

≤0.003% (as SO4)

density

1.670 g/cm3

anion traces

chlorate (ClO3-): ≤10 mg/kg
chloride (Cl-): ≤3 mg/kg
phosphate, silicate (as SiO2): ≤5 mg/kg
sulfate (SO42-): ≤10 mg/kg

cation traces

Ag: ≤0.1 mg/kg
Al: ≤0.05 mg/kg
As: ≤0.05 mg/kg
Ba: ≤0.02 mg/kg
Bi: ≤0.1 mg/kg
Ca: ≤0.5 mg/kg
Cd: ≤0.05 mg/kg
Co: ≤0.05 mg/kg
Cu: ≤0.1 mg/kg
Fe: ≤1 mg/kg
K: ≤0.1 mg/kg
Li: ≤0.02 mg/kg
Mg: ≤0.5 mg/kg
Mn: ≤0.02 mg/kg
Mo: ≤0.05 mg/kg
Ni: ≤0.1 mg/kg
Pb: ≤0.05 mg/kg
Sr: ≤0.02 mg/kg
Tl: ≤0.05 mg/kg
Zn: ≤0.1 mg/kg

SMILES string

OCl(=O)(=O)=O

InChI

1S/ClHO4/c2-1(3,4)5/h(H,2,3,4,5)

InChI key

VLTRZXGMWDSKGL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Perchloric acid (HClO4), an oxygen acid of chlorine, is a colorless liquid. It is strongly acidic in nature. Hydrates of perchloric acid are obtained on its dissolution in water.

Application


  • Development of the Chromatographic Method for Simultaneous Determination of Azaperone and Azaperol in Animal Kidneys and Livers.: This study used perchloric acid for the sample preparation in HPLC analysis, which is crucial for veterinary drug residue analysis in food safety research (Koska and Kubalczyk, 2022).

  • Development of a High Performance Liquid Chromatography with Diode Array Detector (HPLC-DAD) Method for Determination of Biogenic Amines in Ripened Cheeses.: The research utilized perchloric acid for sample deproteinization, demonstrating its application in food chemistry for the quality control of dairy products (Pawul-Gruba et al., 2022).

  • Vibrational and impedance analysis of the beta-stannic acid samples plasticized by perchloric acid.: This study employed perchloric acid in the plasticization process, essential for materials science research on improving the properties of beta-stannic acid (Karelin et al., 2022).

  • Measurement of tigecycline in dried blood spots by LC-MS/MS and comparison tigecycline concentrations between whole blood and plasma.: Perchloric acid was used in the sample preparation for LC-MS/MS analysis, crucial for pharmacokinetic studies in clinical research (Yan et al., 2023).

  • SiO(2)-supported HClO(4) catalyzed synthesis of (Z)-thiazolylhydrazonoindolin-2-ones and their electrochemical properties.: The research highlights the use of perchloric acid as a catalyst, demonstrating its application in organic synthesis and electrochemistry (Shaik et al., 2022).

Other Notes

Reagent for the decomposition of metals and organic compounds (danger of explosions)

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Ox. Liq. 1 - Skin Corr. 1A - STOT RE 2

Target Organs

Thyroid

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Z. Sulcek et al.
Methods of Decomposition in Inorganic Analysis (1989)
R. Bock
Aufschlussmethoden der anorg. & org. Chemie (1972)
Sulfur in plant materials by digestion with nitric and perchloric acid.
Blanchar RW, et al.
Soil Science Society of America Journal. Soil Science Society of America, 29(1), 71-72 (1965)
Eagleson M.
Concise Encyclopedia Chemistry, 790-790 (1994)
Eagleson M.
Concise Encyclopedia Chemistry, 849-849 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service