Skip to Content
Merck
All Photos(1)

Key Documents

71392

Sigma-Aldrich

Sodium chloride solution

technical, ~26% (saturated in water at 20°C, AT)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaCl
CAS Number:
Molecular Weight:
58.44
Beilstein:
3534976
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.55

grade

technical

Quality Level

form

liquid

concentration

~26% (saturated in water at 20°C, AT)

pH

6.5-7.5

SMILES string

[Na+].[Cl-]

InChI

1S/ClH.Na/h1H;/q;+1/p-1

InChI key

FAPWRFPIFSIZLT-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

The product is a saturated solution of sodium chloride in water. Sodium chloride (NaCl) is widely employed preservative for meat, vegetable and fish products. NaCl is a useful precursor for the production of sodium and chlorine containing compounds. Its effect on the growth of salmonellae (mixed inoculum of Salmonella thompson, S. stanley and S. infantis) has been investigated.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michal M Milczarek et al.
Current biology : CB, 28(12), 1975-1980 (2018-06-12)
Memory relies on lasting adaptations of neuronal properties elicited by stimulus-driven plastic changes [1]. The strengthening (and weakening) of synapses results in the establishment of functional ensembles. It is presumed that such ensembles (or engrams) are activated during memory acquisition
Marek Gryta
Chemicke zvesti, 72(1), 99-107 (2018-01-26)
The results of osmotic membrane distillation carried out for 2.5 years were presented in this work. The influence of the process conditions, such as temperature and brine concentration on the permeate flux, was investigated. The saturated NaCl solutions and distilled water
Chentao Yang et al.
BMC genomics, 21(1), 862-862 (2020-12-06)
Over the last decade, the rapid development of high-throughput sequencing platforms has accelerated species description and assisted morphological classification through DNA barcoding. However, the current high-throughput DNA barcoding methods cannot obtain full-length barcode sequences due to read length limitations (e.g.
A M Gibson et al.
International journal of food microbiology, 6(2), 155-178 (1988-03-01)
The growth responses of salmonellae (mixed inoculum of Salmonella thompson, S. stanley and S. infantis) as affected by NaCl concentration, pH level and storage temperature were studied in laboratory medium. Growth curves were obtained at 5 concentrations of NaCl (0.5-4.5%
Eagleson M.
Concise Encyclopedia Chemistry, 996-996 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service