Skip to Content
Merck
All Photos(1)

Key Documents

60235

Sigma-Aldrich

Potassium disulfate

puriss. p.a., ≥97.5% (T)

Synonym(s):

Potassium pyrosulfate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
K2S2O7
CAS Number:
Molecular Weight:
254.32
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21

grade

puriss. p.a.

Quality Level

Assay

≥97.5% (T)

form

powder or crystals

impurities

≤0.002% total nitrogen (N)

pH

1-2 (25 °C, 25.5 g/L)

density

2.28 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤5 mg/kg
phosphate (PO43-): ≤10 mg/kg

cation traces

Ag: ≤5 mg/kg
Al: ≤5 mg/kg
As: ≤5 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Tl: ≤5 mg/kg
Zn: ≤5 mg/kg

SMILES string

[K+].[K+].[O-]S(=O)(=O)OS([O-])(=O)=O

InChI

1S/2K.H2O7S2/c;;1-8(2,3)7-9(4,5)6/h;;(H,1,2,3)(H,4,5,6)/q2*+1;/p-2

InChI key

KAQHZJVQFBJKCK-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Potassium disulfite (Potassium pyrosulfate) has been synthesized from potassium hydrogen sulfate, via melting.

Application

Potassium disulfite supported on silica may be used for the synthesis of ethyl pyruvate.

Other Notes

Reagent for the decomposition of basic metal oxides by fusion

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Eye Dam. 1 - Skin Corr. 1A

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Catalytic Conversion of Diethyl Tartrate into Pyruvate over Silica-Supported Potassium Disulfate.
Sugiyama S, et al.
Bulletin of the Chemical Society of Japan, 65(8), 2083-2085 (1992)
Z. Sulcek et al.
Methods of Decomposition in Inorganic Analysis (1989)
Laura Schneider et al.
Bioresource technology, 234, 1-7 (2017-03-21)
The catalytic conversion of lignocellulosic biomass is attractive due to the feasible generation of valuable products such as reducing sugars which constitute the basic substrates for chemical and transportation fuel production, as well as the production of renewable hydrogen. This
R. Bock
Aufschlussmethoden der anorg. & org. Chemie (1972)
Chromate dermatitis and sodium dithionite.
L M Wall
Contact dermatitis, 8(5), 291-293 (1982-09-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service