Skip to Content
Merck
All Photos(1)

Key Documents

60085

Sigma-Aldrich

Potassium bromate

puriss. p.a., ACS reagent, ≥99.8% (dried material, RT)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KBrO3
CAS Number:
Molecular Weight:
167.00
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NB.24
Assay:
≥99.8% (dried material, RT)
form:
powder or crystals
solubility:
water: soluble(lit.)

grade

ACS reagent
puriss. p.a.

Quality Level

Assay

≥99.8% (dried material, RT)

form

powder or crystals

impurities

≤0.001% total nitrogen (N)

pH

5.0-9.0 (25 °C, 50 mg/mL in H2O)

solubility

water: soluble(lit.)

anion traces

bromide (Br-): ≤200 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Na: ≤100 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

SMILES string

[K+].[O-]Br(=O)=O

InChI

1S/BrHO3.K/c2-1(3)4;/h(H,2,3,4);/q;+1/p-1

InChI key

OCATYIAKPYKMPG-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Potassium bromate (KBrO3) is a colorless solid having trigonal crystals. On heating above 430°C, it degrades and liberates oxygen. It is strongly oxidizing in nature. Its industrial synthesis from potassium bromide via anodic oxidation has been reported. Its quantitative estimation by sensitive flow-injection chemiluminescence method has been proposed. Its standardization has been reported by iodometric titration against sodium thiosulphate (hypo) solution.

Application

Potassium bromate (KBrO3) may be used in volumetric determinations. It may be used as an oxidizing reagent for the oxidation of neomenthol. Kinetics and mechanism of the oxidation reaction has been investigated.

Other Notes

For the in situ production of bromine in TLC; Primary standard for bromometry

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral - Carc. 1B - Ox. Sol. 1

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C. Long, E. & F.N. Spon
Biochemists Handbook, 13-13 (1971)
[ANALYSIS OF DIGITALIS CARDENOLIDES BY THIN LAYER CHROMATOGRAPHY].
L FAUCONNET et al.
Pharmaceutica acta Helvetiae, 38, 423-429 (1963-07-01)
Kinetics and Mechanism of the Oxidation of Neomenthol by Potassium Bromate in Acidic Solution.
Meena R and Sharma S.
International Journal of Pharmacological Research, 4(1), 25-33 (2014)
Chemiluminescence determination of potassium bromate in flour based on flow injection analysis.
Yan Z, et al.
Food Chemistry, 190, 20-24 (2016)
Robert Radford et al.
American journal of physiology. Renal physiology, 302(8), F905-F916 (2012-01-21)
The primary cilium is an immotile sensory and signaling organelle found on the majority of mammalian cell types. Of the multitude of roles that the primary cilium performs, perhaps some of the most important include maintenance of differentiation, quiescence, and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service