Skip to Content
Merck
All Photos(1)

Key Documents

433241

Sigma-Aldrich

Peracetic acid solution

36-40 wt. % in acetic acid

Synonym(s):

Peroxyacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CO3H
CAS Number:
Molecular Weight:
76.05
Beilstein:
1098464
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.21

form

liquid

Quality Level

reaction suitability

reagent type: oxidant

availability

available only in EU

concentration

36-40% (by Na2S2O3, redox titration)
36-40 wt. % in acetic acid

density

1.13 g/cm3

storage temp.

2-8°C

SMILES string

CC(=O)OO

InChI

1S/C2H4O3/c1-2(3)5-4/h4H,1H3

InChI key

KFSLWBXXFJQRDL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Peracetic acid solution may be used as a reaction media to transform phenyldimethylsilyl group to hydroxyl group attached to carbon.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Flam. Liq. 3 - Org. Perox. D - Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.2 - Organic peroxides and self-reacting hazardous materials

WGK

WGK 2

Flash Point(F)

132.8 °F - closed cup

Flash Point(C)

56 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

The Role of Peracetic Acid in Bloodmeal Decoloring.
Hicks TM, et al.
Journal of the American Oil Chemists' Society, 90(10), 1577-1587 (2013)
Peracetic acid oxidation as an alternative pre-treatment for the anaerobic digestion of waste activated sludge.
Appels L, et al.
Bioresource Technology, 102(5), 4124-4130 (2011)
A one-pot conversion of the phenyldimethylsilyl group into a hydroxyl group.
Fleming I and Sanderson PEJ.
Tetrahedron Letters, 28(26), 4229-4232 (1987)
Alberto Baggioli et al.
Physical chemistry chemical physics : PCCP, 15(4), 1130-1140 (2012-12-12)
We examine several computational strategies for the prediction of the (17)O-NMR shielding constants for a selection of organic acids and peracids in aqueous solution. In particular, we consider water (the solvent and reference for the chemical shifts), hydrogen peroxide, acetic
Mehmet Kitis
Environment international, 30(1), 47-55 (2003-12-11)
Peracetic acid is a strong disinfectant with a wide spectrum of antimicrobial activity. Due to its bactericidal, virucidal, fungicidal, and sporicidal effectiveness as demonstrated in various industries, the use of peracetic acid as a disinfectant for wastewater effluents has been

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service