329428
Activated charcoal
Norit® ROW 0.8 SUPRA, pellets
Synonym(s):
Charcoal activated
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
C
CAS Number:
Molecular Weight:
12.01
EC Number:
MDL number:
UNSPSC Code:
11101522
PubChem Substance ID:
NACRES:
SB.52
Recommended Products
vapor pressure
<0.1 mmHg ( 20 °C)
Quality Level
form
pellets
autoignition temp.
842 °F
manufacturer/tradename
Norit® ROW 0.8 SUPRA
greener alternative product characteristics
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
resistivity
1375 μΩ-cm, 20°C (graphite)
mp
3550 °C (lit.)
greener alternative category
, Enabling
SMILES string
[C]
InChI
1S/C
InChI key
OKTJSMMVPCPJKN-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Click here for more details.
Application
Activated charcoal (NORIT® ROW 0.8 SUPRA) can be used as an adsorbent for the adsorption of phenol in aqueous solution. It can also be used in the adsorption of fulvic acid for potential usage in the treatment of waste water.
Legal Information
Norit is a registered trademark of Norit N.V.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Effect of iron and aluminum on adsorption of fulvic acids on Norit ROW 0.8 Supra carbon
Grzegorczuk-Nowacka M and Anielak AM
Environmental Engineering Science, 34(9), 659-665 (2017)
Improved adsorption capacity of commercially available activated carbon norit ROW 0.8 Supra with thermal treatment for phenol removal
Citraningrum M, et al.
Journal of Environmental Protection, 1, 62-74 (2007)
Activated carbon for gas separation and storage
Sircar S, et al.
Carbon, 34(1), 1-12 (1996)
About reactions occurring during chemical activation with hydroxides.
Lillo-Rodenas MA, et al.
Carbon, 42(7), 1371-1375 (2004)
Adsorption of dyes from aqueous solutions on activated charcoal.
Iqbal MJ, et al.
Journal of Hazardous Materials, 139(1), 57-66 (2007)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service