Skip to Content
Merck
All Photos(1)

Key Documents

32304

Sigma-Aldrich

Ammonium oxalate monohydrate

puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., 99.5-101.0%

Synonym(s):

Oxalic acid diammonium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NH4)2C2O4 · H2O
CAS Number:
Molecular Weight:
142.11
Beilstein:
3759830
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
Assay:
99.5-101.0%
form:
crystalline
Pricing and availability is not currently available.

grade

ACS reagent
puriss. p.a.

Quality Level

Agency

USP/NF
reag. ISO
reag. Ph. Eur.

vapor pressure

<0.1 hPa ( 20 °C)

Assay

99.5-101.0%

form

crystalline

impurities

≤0.005% in water insoluble matters

ign. residue

≤0.02% (as SO4)

pH

6-7 (20 °C, 2.5%)

mp

133 °C (dec.) (lit.)

anion traces

chloride (Cl-): ≤5 mg/kg
nitrate (NO3-): ≤5 mg/kg
sulfate (SO42-): ≤20 mg/kg

cation traces

Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤2 mg/kg
K: ≤10 mg/kg
Mg: ≤10 mg/kg
Na: ≤10 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

SMILES string

OC(C(O)=O)=O.N.O.N

InChI

1S/C2H2O4.2H3N.H2O/c3-1(4)2(5)6;;;/h(H,3,4)(H,5,6);2*1H3;1H2

InChI key

MSMNVXKYCPHLLN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Ammonium oxalate monohydrate is the diammonium salt of oxalic acid. Its refined crystal structure has been investigated by three-dimensional diffraction studies.[1] Influence of bivalent [Mn(II), Co(II), Ni(II)] and trivalent cations [Fe(III), Cr(III)] impurities on the growth rates of ammonium oxalate monohydrate crystals has been described.[2] Its space group and unit cell parameters have been analyzed by neutron diffraction studies.[3] Its IR spectra has been compared with that of isolated coal-solubilizing agent (CSA).[4]

Application

Ammonium oxalate monohydrate may be used to prepare an electrochemiluminescence (ECL) solution containing 1mM tris(2,2-bipyridyl)ruthenium(II) (Ru(bpy)32+) and 0.2mM ammonium oxalate monohydrate in a 0.1M ammonium acetate buffer at pH 5.0.[5]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M S Cohen et al.
Applied and environmental microbiology, 56(11), 3285-3291 (1990-11-01)
Low-ranked coals were dissolved by using cell extracts derived from liquid cultures of Trametes versicolor. The coal-solubilizing agent (CSA) was separated from the broth components by a multistep isolation procedure including reverse-phase high-pressure liquid chromatography, size exclusion chromatography, ethanol fractionation
Neutron diffraction study of ammonium oxalate monohydrate,(NH4)2C2O4.H2O.
Padmanabhan VM, et al.
Acta Crystallographica, 18(3), 567-568 (1965)
Ammonium oxalate monohydrate: structure refinement at 30K.
Robertson JH.
Acta Crystallographica, 18(3), 410-417 (1965)
Effect of impurities on metastable zone width for the growth of ammonium oxalate monohydrate crystals from aqueous solutions.
Sangwal K and Mielniczek-Brzoska E.
Journal of Crystal Growth, 267(3), 662-675 (2004)
Electrochemiluminescence as a tool for microscopy at the nanoscale.
Lee A, et al.
Proceedings of SPIE, 13, 13-20 (2004)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service