Skip to Content
Merck
All Photos(1)

Key Documents

31633

Sigma-Aldrich

Strontium nitrate

puriss. p.a., ACS reagent, ≥99.0% (complexometric)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Sr(NO3)2
CAS Number:
Molecular Weight:
211.63
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NB.24

grade

ACS reagent
puriss. p.a.

Assay

≥99.0% (complexometric)

form

powder or crystals

impurities

≤0.005% ammonium (NH4)
≤0.01% in water insoluble matters (10% solution)
≤0.15% Mg and alkali salts
≤5 ppm heavy metals (by ICP-OES)

loss

≤0.1% loss on drying, 105 °C (4h)

pH

5.0-7.0 (25 °C, 5%)

mp

570 °C (lit.)

anion traces

chloride (Cl-): ≤10 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Ba: ≤100 mg/kg
Ca: ≤50 mg/kg
Cu: ≤2 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Mg: ≤20 mg/kg
Na: ≤50 mg/kg
Zn: ≤2 mg/kg

SMILES string

[Sr++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/2NO3.Sr/c2*2-1(3)4;/q2*-1;+2

InChI key

DHEQXMRUPNDRPG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Strontium nitrate is a salt of strontium. Its crystal structure has been studied by neutron diffraction. Crystals of strontium nitrate have been reported to be cubic.

Application

Strontium nitrate may be used in the preparation of La0.4Sr0.5Ba0.1TiO3 (LSBT) precursor solution.

Pictograms

Flame over circleCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Ox. Sol. 1

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G S Hahn
Dermatologic surgery : official publication for American Society for Dermatologic Surgery [et al.], 25(9), 689-694 (1999-09-22)
Irritant contact dermatitis occurs following topical exposure to many chemicals found in cosmetics, personal care products, drugs, and during occupational exposure. Signs and symptoms may include sensory irritation (sting, burn, and/or itch), erythema, edema, and vesiculation. In an attempt to
Porous YSZ impregnated with La0.4Sr0.5Ba0.1TiO3 as a possible composite anode for SOFCs fueled with sour feeds.
Vincent AL, et al.
Journal of Power Sources, 215, 301-306 (2012)
H Zhai et al.
Dermatology (Basel, Switzerland), 200(3), 244-246 (2000-06-01)
Previous studies have demonstrated that strontium salts have a potent and broad ability to suppress sensory irritation. To ascertain the possible antipruritic effects of topical strontium salts, we conducted a double-blind randomized study to evaluate the effect of 20% strontium
H Zhai et al.
Contact dermatitis, 42(2), 98-100 (2000-03-07)
Skin care products are complex formulations that may cause sensory irritation symptoms, characterized by stinging, burning, and itching. Substances capable of counteracting sensory irritation are of great practical interest. Strontium salts have been demonstrated to inhibit sensory irritation and inflammation
Sangita Pandey et al.
Journal of magnetic resonance (San Diego, Calif. : 1997), 209(2), 220-226 (2011-02-08)
Electron paramagnetic resonance (EPR) study of Fe³(+) ions doped strontium nitrate (SN) single crystals is performed at liquid nitrogen temperature and at X band frequency. The spin Hamiltonian (SH) parameters are determined from the resonance lines observed at different angular

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service