Skip to Content
Merck
All Photos(2)

Key Documents

211907

Sigma-Aldrich

Lead(II) oxide

powder, <10 μm, ReagentPlus®, ≥99.9% trace metals basis

Synonym(s):

Lead monoxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
PbO
CAS Number:
Molecular Weight:
223.20
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.55
Assay:
≥99.9% trace metals basis
form:
powder
Pricing and availability is not currently available.

vapor pressure

10 mmHg ( 0 °C)

Quality Level

product line

ReagentPlus®

Assay

≥99.9% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: lead

particle size

<10 μm

pH

9.9 (20 °C, 100 g/L)

mp

886 °C (lit.)

SMILES string

O=[PbH2]

InChI

1S/O.Pb

InChI key

YEXPOXQUZXUXJW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Lead (II) oxide is amphoteric in nature, capable of reacting with both acid and base.[1] It exists in two forms, litharge and massicot.[2] Lead (II) oxide is commonly used in making lead glass and glazing pottery.[3] Addition of PbO (as a dopant) to Ni-Cu-Zn ferrite increases the densification rate and final sintered density, whereas activation energy of densification was reduced.[4]

Application

Lead(II) oxide (PbO) has been used in the preparation of (Pb0.93La0.07)(Zr0.65Ti0.35)O3 powder (PLZT 7/65/35),[5] (Pb0.97La0.03)(Zr0.52Ti0.48)O3 (PLZT) ceramics[6] and (Pb0.84Ba0.16)(Zr0.54Ti0.46)O3 (PBZT) ceramics.[6]

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Lact. - Repr. 1A - STOT RE 1

Target Organs

Central nervous system,Kidney,Blood

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Magnetic and piezoelectric properties of magnetoelectric laminate transformer.
Kozielski L and Adamczyk M.
Archives of Metallurgy and Materials, 56(4), 1111-1117 (2011)
Clugston M and Flemming R
Advanced Chemistry, 340-340 (2000)
Effects of Lead (II) Oxide on Processing and Properties of Low-Temperature-Cofirable Ni-Cu-Zn Ferrite.
Jean JH, et al.
Journal of the American Ceramic Society. American Ceramic Society, 82(2), 343-350 (1999)
Daintith J.
The facts on file dictionary of chemistry, 161-161 (1999)
Lucjan Kozielski et al.
Sensors (Basel, Switzerland), 13(2), 2419-2429 (2013-02-14)
Enhanced functionality of electro-optic devices by implementing piezoelectric micro fibers into their construction is proposed. Lanthanum-modified lead zirconate titanate (PLZT) ceramics are known to exhibit high light transparency, desirable electro-optic properties and fast response. In this study PLZT fibers

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service