All Photos(1)
About This Item
Linear Formula:
Na86[AlO2)86(SiO2)106] · xH2O
CAS Number:
MDL number:
UNSPSC Code:
23201100
eCl@ss:
32110204
NACRES:
SB.52
Recommended Products
form
beads
particle size
8-12 mesh
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Molecular sieves are crystalline metal aluminosilicates having a three-dimensional interconnecting network of silica and alumina tetrahedra. Natural water of hydration is removed from this network by heating to produce uniform cavities which selectively adsorb molecules of a specific size. This sodium form represents the basic structure of the type X family, with an effective pore opening in the 910¼ range (for example: it will not adsorb perfluorotributylamine). The 8 to 12-mesh type is common in liquid-phase applications.
Molecular sieves, 13X showed preferential adsorption of CO, over nitrogen or hydrogen at all pressures up to 250psia.
Application
Molecular sieves, 13X may be used for commercial gas drying, air plant-feed purification (simultaneous water and carbon dioxide removal) and liquid hydrocarbon/natural gas sweetening (hydrogen sulfide and mercaptan removal).
The product (Molecular sieves, 13X) have been used for the abstraction of water from the 100% ethanol.
Also, they are widely employed as an adsorbent in adsorption studies.
Also, they are widely employed as an adsorbent in adsorption studies.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Victoria C Cogger et al.
Journal of visualized experiments : JoVE, (98)(98), e52698-e52698 (2015-05-21)
Liver sinusoidal endothelial cells are the gateway to the liver, their transcellular fenestrations allow the unimpeded transfer of small and dissolved substances from the blood into the liver parenchyma for metabolism and processing. Fenestrations are dynamic structures--both their size and/or
Adsorption of CO2 on molecular sieves and activated carbon.
Siriwardane RV, et al.
Energy and Fuels, 15(2), 279-284 (2001)
Equilibrium adsorption of ethane, ethylene, isobutane, carbon dioxide, and their binary mixtures on 13X molecular sieves.
Hyun SH and Danner RP.
Journal of Chemical and Engineering Data, 27(2), 196-200 (1982)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service