Skip to Content
Merck
All Photos(1)

Key Documents

12658

Sigma-Aldrich

Potassium sulfate

meets analytical specification of Ph. Eur., 99-101%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K2SO4
CAS Number:
Molecular Weight:
174.26
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

Quality Level

Assay

99-101%

form

solid

quality

meets analytical specification of Ph. Eur.

impurities

acidity or alkalinity, in accordance
residual solvents, complies
≤0.001% heavy metals (as Pb)

loss

≤0.5% loss on drying, 130 °C

solubility

boiling water: soluble 1 gm in 4 ml
glycerol: soluble 1 gm in 75 ml
water: soluble 1 gm in 8.3 ml
alcohol: insoluble

anion traces

chloride (Cl-): ≤20 mg/kg

cation traces

As: ≤2 mg/kg
Ca: ≤50 mg/kg
Fe: ≤10 mg/kg
Mg: ≤20 mg/kg
Na: ≤100 mg/kg

suitability

in accordance for appearance of solution

SMILES string

[K+].[K+].[O-]S([O-])(=O)=O

InChI

1S/2K.H2O4S/c;;1-5(2,3)4/h;;(H2,1,2,3,4)/q2*+1;/p-2

InChI key

OTYBMLCTZGSZBG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael Thompson et al.
The Analyst, 127(12), 1666-1668 (2003-01-23)
Both the Kjeldahl and the Dumas methods for the determination of protein in foodstuffs are currently in use, but the empirical nitrogen factors used to convert the determined nitrogen content to protein content are based on the Kjeldahl method alone.
T L Little et al.
Circulation research, 76(3), 498-504 (1995-03-01)
Dye tracers were chosen, based on net charge, chemical structure, and reactive groups, to test for the existence of and to provide novel insight into channel selectivities of junctional pathways connecting smooth muscle and endothelial cells of the arteriolar wall.
S S Lehrer et al.
Journal of structural biology, 122(1-2), 176-179 (1998-09-02)
The alpha-helical coiled-coil tropomyosin homodimer, alpha alphaTm, unfolds cooperatively with T1/2 = 47 degreesC, at neutral pH, 0.5 M NaCl. At pH 2, where each chain contains 55 positive charges, no cooperative unfolding occurs to at least 80 degreesC. The
Jinyong Yan et al.
Bioresource technology, 102(7), 4755-4758 (2011-02-01)
A dual modification procedure composed of cross-linking and protein coating with K(2)SO(4) was employed to modify Geotrichum sp. lipase for catalyzing biodiesel production from waste cooking oil. Compared to single modification of protein coating with K(2)SO(4), the dual modification of
Ashley C Parker et al.
Clinical orthopaedics and related research, 469(11), 3008-3015 (2011-05-10)
Local drug delivery has substantial potential to prevent infections compared with systemic delivery. Although calcium sulfate (CaSO(4)) has been studied for local drug delivery and two types are commercially available, it is unknown whether they differentially release antibiotics. We determined

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service