Skip to Content
Merck
All Photos(2)

Key Documents

125008

Sigma-Aldrich

1-Chlorobutane

ReagentPlus®, 99%

Synonym(s):

Butyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3Cl
CAS Number:
Molecular Weight:
92.57
Beilstein:
1730909
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.04

vapor density

3.2 (vs air)

Quality Level

vapor pressure

80.1 mmHg ( 78.4 °C)

product line

ReagentPlus®

Assay

99%

form

liquid

autoignition temp.

860 °F

expl. lim.

10.1 %

refractive index

n20/D 1.402 (lit.)

bp

77-78 °C (lit.)

mp

−123 °C (lit.)

solubility

water: soluble 0.11 g/L at 20 °C

density

0.886 g/mL at 25 °C (lit.)

SMILES string

CCCCCl

InChI

1S/C4H9Cl/c1-2-3-4-5/h2-4H2,1H3

InChI key

VFWCMGCRMGJXDK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Chlorobutane is an alkyl halide. It undergoes reaction with nanocrystalline MgO at 200-350°C to afford butane isomers and MgCl2, via dehydrochlorination.

Application

1-Chlorobutane may be used in the synthesis of ionic liquids, 1-butyl-3-methylimidazolium hydrogen sulfate ([Bmim]+[HSO4]-) and 1-butyl-3-methylimidazolium dihydrogen phosphate ([Bmim]+[H2PO4]-).

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

10.4 °F - closed cup

Flash Point(C)

-12 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mannich reaction using acidic ionic liquids as catalysts and solvents.
Zhao G, et al.
Green Chemistry, 6(2), 75-77 (2004)
Changes in texture and catalytic activity of nanocrystalline MgO during its transformation to MgCl2 in the reaction with 1-chlorobutane.
Fenelonov VB, et al.
The Journal of Physical Chemistry B, 105(18), 3937-3941 (2001)
Virendra Patil et al.
Chemosphere, 243, 125302-125302 (2019-11-15)
Four 1-butyl-3-methylimidazolium halide ionic liquids were synthesized via metathesis and anion exchange reactions. Silver nanoparticles (AgNPs) colloids were synthesized in four ionic liquids in the pressurized reactor by reduction of silver nitrate with hydrogen gas, without adding solvents or stabilizing
Bing-Jyun Lu et al.
Scientific reports, 9(1), 13253-13253 (2019-09-15)
Wood has been a crucial natural material for human civilization since prehistoric times. In archaeology, the examination of the wood microstructure is important for the study of architecture, musical instruments, sculptures, and so on. Scanning electron microscopy (SEM) examination is
Secil Öztürk et al.
Beilstein journal of nanotechnology, 11, 770-781 (2020-06-09)
Covalent triazine frameworks (CTFs) are little investigated, albeit they are promising candidates for electrocatalysis, especially for the oxygen evolution reaction (OER). In this work, nickel nanoparticles (from Ni(COD)2) were supported on CTF-1 materials, which were synthesized from 1,4-dicyanobenzene at 400

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service