Skip to Content
Merck
All Photos(1)

Key Documents

12010

Sigma-Aldrich

Calcium carbonate

puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E170, precipitated, 98.5-100.5% (based on anhydrous substance)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CaCO3
CAS Number:
Molecular Weight:
100.09
Beilstein:
8008338
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.21

grade

puriss.

Quality Level

Assay

98.5-100.5% (based on anhydrous substance)

form

powder

quality

meets analytical specification of Ph. Eur., BP, USP, FCC, E170
precipitated

impurities

organic volatile impurities, complies
residual solvents, complies
≤0.002% heavy metals (as Pb)
≤0.2% insoluble in acid (CH3COOH)
≤0.2% insoluble in hydrochloric acid
≤1% alkali-, Mg-ions (as SO4)

loss

≤2% loss on drying, 200 °C

pH

8

density

2.93 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤200 mg/kg
fluoride (F-): ≤50 mg/kg
sulfate (SO42-): ≤2000 mg/kg

cation traces

As: ≤2 mg/kg
Ba:, in accordance
Cu: ≤10 mg/kg
Fe: ≤200 mg/kg
Hg: ≤0.5 mg/kg
Pb: ≤3 mg/kg
Zn: ≤10 mg/kg

SMILES string

[Ca++].[O-]C([O-])=O

InChI

1S/CH2O3.Ca/c2-1(3)4;/h(H2,2,3,4);/q;+2/p-2

InChI key

VTYYLEPIZMXCLO-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application


  • Heavy metal immobilization and radish growth improvement using Ca(OH)(2)-treated cypress biochar in contaminated soil.: This study demonstrates the efficacy of Ca(OH)(2)-treated cypress biochar, rich in calcium carbonate, in immobilizing heavy metals and enhancing radish growth in contaminated soils, offering sustainable solutions for soil remediation (Shah et al., 2024).

  • Liming potential and characteristics of biochar produced from woody and non-woody biomass at different pyrolysis temperatures.: This research investigates the liming potential of biochar, particularly focusing on the role of calcium carbonate in neutralizing soil acidity and improving soil health, thus promoting sustainable agricultural practices (Murtaza et al., 2024).

  • Valorisation diagnosis of waste from the decontamination of phosphogypsum leachates through a combined calcium carbonate/hydroxide process.: This study evaluates the effectiveness of a combined calcium carbonate/hydroxide process in decontaminating phosphogypsum leachates, highlighting the potential of this method in waste valorization and environmental management (Soto-Cruz et al., 2024).


Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yves F Widmer et al.
eLife, 7 (2018-10-23)
Lasting changes in gene expression are critical for the formation of long-term memories (LTMs), depending on the conserved CrebB transcriptional activator. While requirement of distinct neurons in defined circuits for different learning and memory phases have been studied in detail
Sophie A Jamal et al.
Lancet (London, England), 382(9900), 1268-1277 (2013-07-23)
Phosphate binders (calcium-based and calcium-free) are recommended to lower serum phosphate and prevent hyperphosphataemia in patients with chronic kidney disease, but their effects on mortality and cardiovascular outcomes are unknown. We aimed to update our meta-analysis on the effect of
Polysaccharides and proteoglycans in calcium carbonate-based biomineralization.
José L Arias et al.
Chemical reviews, 108(11), 4475-4482 (2008-07-26)
Ling Li et al.
Nature materials, 13(5), 501-507 (2014-04-01)
Hierarchical composite materials design in biological exoskeletons achieves penetration resistance through a variety of energy-dissipating mechanisms while simultaneously balancing the need for damage localization to avoid compromising the mechanical integrity of the entire structure and to maintain multi-hit capability. Here
Claudia Gagnon et al.
PloS one, 9(10), e109607-e109607 (2014-10-10)
To examine whether combined vitamin D and calcium supplementation improves insulin sensitivity, insulin secretion, β-cell function, inflammation and metabolic markers. 6-month randomized, placebo-controlled trial. Ninety-five adults with serum 25-hydroxyvitamin D [25(OH)D] ≤55 nmol/L at risk of type 2 diabetes (with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service