Skip to Content
Merck
All Photos(1)

Key Documents

11044

Sigma-Aldrich

Aluminum sulfate octadecahydrate

puriss., meets analytical specification of Ph. Eur., BP, 100-110%, 51.0-59.0% Al2(SO4)3 basis

Synonym(s):

Aluminum sulfate hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Al2(SO4)3 · 18H2O
CAS Number:
Molecular Weight:
666.43
EC Number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21

Quality Level

grade

puriss.

Assay

100-110%

form

crystals or chunks

quality

meets analytical specification of Ph. Eur., BP

concentration

51.0-59.0% Al2(SO4)3

impurities

residual solvents, complies
≤0.002% heavy metals (as Pb)
≤0.003% ammonium (NH4)
≤0.2% sub.not precip.b.NH4OH(SO4)

pH

2.5-4.0 (20 °C, 2%)

anion traces

chloride (Cl-): ≤50 mg/kg

cation traces

As: ≤3 mg/kg
Fe: ≤50 mg/kg

suitability

complies for appearance of solution

SMILES string

O=S(O)(O)=O.[AlH2][AlH2].O

InChI

1S/2Al.3H2O4S.9H2O/c;;3*1-5(2,3)4;;;;;;;;;/h;;3*(H2,1,2,3,4);9*1H2/q2*+3;;;;;;;;;;;;/p-6

InChI key

KFCWRPFNNIEZRY-UHFFFAOYSA-H

Looking for similar products? Visit Product Comparison Guide

General description

Aluminum sulfate octadecahydrate (Al2(SO4)3.18H2O) is a hydrated aluminum salt. It is used as a catalyst and cross-linker during chemical reactions. Aluminum sulfate octadecahydrate is also used as a coagulating agent/flocculating agent in the purification of drinking water and wastewater treatment plants. It is also used in the paper industry for sizing.

Application

Aluminum sulfate octadecahydrate is used:
  • As a trace water source in the synthesis of cancrinite zeolite from geophagic clay.
  • In the solvent free synthesis of ZSM-5 Zeolites from rice husk silica.
  • In the preparation of polymeric metal complex dyes.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Nak Cheon Jeong et al.
Langmuir : the ACS journal of surfaces and colloids, 21(13), 6038-6047 (2005-06-15)
Various MX (M = Cd, Zn, and Mn, X = S and Se) semiconductor quantum dots (QDs) were prepared in zeolite Y. While the QDs are readily expelled from zeolite interior upon exposure of the MX QD-incorporating zeolite Y ([MX]-Y)
Hydrolysis products of water treatment chemical aluminium sulfate octadecahydrate by electrospray ionization mass spectrometry.
Sarpola A, et al.
Polyhedron, 26(12), 2851-2858 (2007)
Synthesis and light emitting properties of polymeric metal complex dyes based on hydroxyquinoline moiety.
Kim S-H, et al.
Dyes and Pigments, 55(2), 91-97 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service