Skip to Content
Merck
All Photos(1)

Key Documents

11019

Sigma-Aldrich

Aluminum chloride

anhydrous, sublimed, ≥98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
AlCl3
CAS Number:
Molecular Weight:
133.34
EC Number:
MDL number:
UNSPSC Code:
12352302
eCl@ss:
38120507
PubChem Substance ID:
NACRES:
NA.21
Assay:
≥98%
form:
powder
Pricing and availability is not currently available.

vapor pressure

1 mmHg ( 100 °C)

Quality Level

Assay

≥98%

form

powder

quality

anhydrous
sublimed

reaction suitability

reagent type: catalyst
core: aluminum

impurities

≤0.005% heavy metals (as Pb)

pH

2.4 (20 °C, 100 g/L)

mp

190 °C (lit.)

anion traces

sulfate (SO42-): ≤500 mg/kg

cation traces

Fe: ≤200 mg/kg

SMILES string

Cl[Al](Cl)Cl

InChI

1S/Al.3ClH/h;3*1H/q+3;;;/p-3

InChI key

VSCWAEJMTAWNJL-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Application


  • The coagulation process for virus removal in turbid water: Explores the use of aluminum chloride in the coagulation process to remove both enveloped and non-enveloped viruses, including SARS-CoV-2 Omicron BA.2, from turbid water. This research is crucial for environmental lab technicians and researchers in academia focusing on water treatment and public health safety (Maneein S et al., 2024).

  • Enhancing pollutants removal in hospital wastewater: Compares PAC coagulation and bio-contact oxidation methods in wastewater treatment, underscoring the impact of outdated plants and the role of aluminum chloride in enhancing pollutant removal efficiency. This study is essential for environmental scientists and engineers tasked with upgrading and optimizing wastewater treatment processes (Kang Y et al., 2024).

  • Comparing the effects of Al-based coagulants in waste activated sludge anaerobic digestion: Analyzes methane yield, kinetics, and sludge implications when using aluminum-based coagulants, providing valuable insights for environmental engineers and material scientists interested in resource recovery and sustainable waste management (Pasciucco F et al., 2024).

Features and Benefits

The vapor-phase co-reductions with other metal halides by hydrogen results in finely divided intermetallics with applications as structural materials or compounds with useful thermoelectric, magnetic, and oxidation-resistance properties. Aluminum nitride can be synthesized from the elemnts with AlCl3 added to help facilitate crystallization.

Caution

Caution when opening! Due to production the supernatant gas phase can contain small amounts of toxic phosgene.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hattori H, et al.
Acid-Base Catalysis II (1994)
Spectroscopy and electrochemistry of uranium (IV)/uranium (III) in basic aluminum chloride-1-ethyl-3-methylimidazolium chloride.
Anderson CJ, et al.
Inorganic Chemistry, 30(21), 4013-4016 (1991)
THE FRIES REARRANGEMENT OF PHENYL ISOBUTYRATE.
Briggs TI, et al.
Canadian Journal of Chemistry, 34(7), 851-855 (1956)
Nemoto T, et al.
Polymer Journal, 40(7) (2008)
Amination of< i> exo</i>-2-chloronorbornane and norbornane with trichloramine-aluminum chloride.
Kovacic P, et al.
Tetrahedron, 26(2), 529-538 (1970)

Questions

  1. Can this product be used in total flavonoid content assays?

    1 answer
    1. This product has not been evaluated for total flavonoid assays. Validation for this application will be the responsibility of the end user. Published literature suggests it may be suitable for this purpose. Please see the citation below for further information on this type of an application.

      Yagi, S.; Nilofar; Zengin, G.; Yildiztugay, E.; Caprioli, G.; Piatti, D.; Menghini, L.; Ferrante, C.; Di Simone, S.C.; Chiavaroli, A.; et al. Exploring for HPLC-MS/MS Profiles and Biological Activities of Different Extracts from Allium lycaonicum Siehe ex Hayek from Turkey Flora. Foods 2023, 12, 4507. https://doi.org/10.3390/foods12244507

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service