Key Documents
Z515639
Sigma-Aldrich
Aldrich® evaporating flask, pear-shaped
capacity 1,000 mL, joint: ST/NS 24/40, non-coated, heavy wall
Synonym(s):
bottom flask, flask round, round flask
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
Pricing and availability is not currently available.
material
borosilicate glass
long
pear-shaped
single-neck
Joint
joint: ST/NS24/40
sterility
non-sterile
feature
closure type Standard Taper Joint
packaging
pack of 1
manufacturer/tradename
Büchi equivalent, 00440
capacity
1,000 mL
Compare Similar Items
View Full Comparison
Show Differences
1 of 4
This Item
Z515647Z515574Z515582
material
borosilicate glass , pear-shaped , long , single-neck
material
borosilicate glass , long , pear-shaped , single-neck
material
borosilicate glass , long , pear-shaped , single-neck
material
borosilicate glass , long , pear-shaped , single-neck
manufacturer/tradename
Büchi equivalent, 00440
manufacturer/tradename
Büchi equivalent, 00435
manufacturer/tradename
Büchi equivalent, 08751
manufacturer/tradename
Büchi equivalent, 00432
joint
joint: ST/NS24/40
joint
joint: ST/NS29/32
joint
joint: ST/NS24/40
joint
joint: ST/NS29/32
packaging
pack of 1
packaging
pack of 1
packaging
pack of 1
packaging
pack of 1
sterility
non-sterile
sterility
non-sterile
sterility
non-sterile
sterility
non-sterile
General description
These pear-shaped evaporating flasks can be used as boiling type flasks for rotary evaporators.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)