Y0001589
Atomoxetine impurity D
European Pharmacopoeia (EP) Reference Standard
About This Item
Recommended Products
grade
pharmaceutical primary standard
API family
atomoxetine
manufacturer/tradename
EDQM
application(s)
pharmaceutical (small molecule)
format
neat
storage temp.
2-8°C
1 of 4
This Item | Y0001588 | Y0001587 | Y0001571 |
---|---|---|---|
pharmaceutical primary standard | pharmaceutical primary standard | pharmaceutical primary standard | pharmaceutical primary standard |
pharmaceutical (small molecule) | pharmaceutical (small molecule) | pharmaceutical (small molecule) | pharmaceutical (small molecule) |
neat | neat | neat | neat |
atomoxetine | atomoxetine | atomoxetine | naloxone |
2-8°C | 2-8°C | 2-8°C | 2-8°C |
General description
Application
Packaging
Other Notes
related product
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
215.6 °F
Flash Point(C)
102 °C
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service