Skip to Content
Merck
All Photos(1)

Key Documents

Y0001445

Celecoxib

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

4-[5-(4-Methylphenyl)-3-(trifluoromethyl)-1H-pyrazol-1-yl]benzenesulfonamide, CJ-016377, CP-598107, PF-00345549, PHA-00846533, SC-58635, YM-177

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H14F3N3O2S
CAS Number:
Molecular Weight:
381.37
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

celecoxib

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Cc1ccc(cc1)-c2cc(nn2-c3ccc(cc3)S(N)(=O)=O)C(F)(F)F

InChI

1S/C17H14F3N3O2S/c1-11-2-4-12(5-3-11)15-10-16(17(18,19)20)22-23(15)13-6-8-14(9-7-13)26(21,24)25/h2-10H,1H3,(H2,21,24,25)

InChI key

RZEKVGVHFLEQIL-UHFFFAOYSA-N

Gene Information

human ... PTGS2(5743)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Celecoxib EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Celecoxib is a non-steroidal, anti-inflammatory drug (NSAID) and a cyclooxygenase-2 (COX-2) selective inhibitor. Celecoxib is at least 10-20 times more selective for COX-2 over COX-1.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 1 - Repr. 1B - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Verena Jendrossek
Cancer letters, 332(2), 313-324 (2011-02-25)
Celecoxib is a paradigmatic selective inhibitor of cyclooxygenase-2 (COX-2). This anti-inflammatory drug has potent anti-tumor activity in a wide variety of human epithelial tumor types, such as colorectal, breast, non-small cell lung, and prostate cancers. Up to now, the drug
Axel H Schönthal et al.
Expert opinion on investigational drugs, 17(2), 197-208 (2008-01-31)
Celecoxib is an NSAID that was developed as a selective inhibitor of COX-2 and approved by the FDA for the treatment of various forms of arthritis and the management of acute or chronic pain. In addition, it was more recently
Sheena Derry et al.
The Cochrane database of systematic reviews, 10(10), CD004233-CD004233 (2013-10-24)
This is an update of a review first published in The Cochrane Library in Issue 4, 2008, and updated in Issue 3, 2012. Celecoxib is a selective cyclo-oxygenase-2 (COX-2) inhibitor usually prescribed for the relief of chronic pain in osteoarthritis
Paul L McCormack
Drugs, 71(18), 2457-2489 (2011-12-07)
Celecoxib (Celebrex®) was the first cyclo-oxygenase (COX)-2 selective inhibitor (coxib) to be introduced into clinical practice. Coxibs were developed to provide anti-inflammatory/analgesic activity similar to that of nonselective NSAIDs, but without their upper gastrointestinal (GI) toxicity, which is thought to
Leyte L Winfield et al.
Future medicinal chemistry, 4(3), 361-383 (2012-03-08)
Celecoxib is a multifaceted drug with promising anticancer properties. A number of studies have been conducted that implicate the compound in modulating the expression of Bcl-2 family members and mitochondria-mediated apoptosis. The growing data surrounding the role of celecoxib in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service