Skip to Content
Merck
All Photos(1)

Key Documents

Y0001297

Iopentol

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

5-[Acetyl(2-hydroxy-3-methylpropyl)amino]-N,N′-bis(2,3-dihydroxypropyl)-2,4,6-triiodo-1,3-benzenedicarboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H28I3N3O9
CAS Number:
Molecular Weight:
835.16
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

iopentol

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Ic1c(c(c(c(c1C(=O)NCC(O)CO)I)C(=O)NCC(O)CO)I)N(CC(O)COC)C(=O)C

InChI

1S/C20H28I3N3O9/c1-9(29)26(5-12(32)8-35-2)18-16(22)13(19(33)24-3-10(30)6-27)15(21)14(17(18)23)20(34)25-4-11(31)7-28/h10-12,27-28,30-32H,3-8H2,1-2H3,(H,24,33)(H,25,34)

InChI key

IUNJANQVIJDFTQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Iopentol EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Steffen Ross et al.
AJR. American journal of roentgenology, 190(5), 1380-1389 (2008-04-24)
The objective of our study was to establish a standardized procedure for postmortem whole-body CT-based angiography with lipophilic and hydrophilic contrast media solutions and to compare the results of these two methods. Minimally invasive postmortem CT angiography was performed on
P Dewachter et al.
Journal de radiologie, 82(9 Pt 1), 973-977 (2001-10-10)
The etiology of severe reactions following injection of iodinated contrast agents is the subject of controversy. No consensus has been established regarding the management of patients at risk, risk factors and premedication because in most cases published no diagnostic exploration
K Katsanos et al.
Clinical hemorheology and microcirculation, 39(1-4), 87-91 (2008-05-27)
Iodinated contrast media (CM) are widely used in diagnostic imaging and therapeutic interventional procedures of everyday clinical practice and are associated with multiple hemodynamic and hemorheological effects. The purpose of our work was to investigate the red blood cell (RBC)
J E Roos et al.
RoFo : Fortschritte auf dem Gebiete der Rontgenstrahlen und der Nuklearmedizin, 176(4), 556-563 (2004-04-17)
To evaluate the impact of different iodine concentrations of intravenous contrast agent on hepatic and vascular enhancement during arterial and porto-venous phase imaging using a 4-channel multi-detector row CT (MDCT). One hundred consecutive patients referred for triphasic abdominal MDCT were
G Jung et al.
Journal of computer assisted tomography, 24(5), 783-787 (2000-10-25)
To evaluate the diagnostic value of MR imaging in abdominal lymphoma staging, in comparison with that of the established spiral CT, within the context of a prospective study. 50 patients with non-Hodgkin (n = 27) and Hodgkin lymphoma (n =

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service