Skip to Content
Merck
All Photos(1)

Key Documents

Y0001284

Lufenuron

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

1-[2,5-Dichloro-4-(1,1,2,3,3,3-hexafluoropropoxy)phenyl]-3-(2,6-difluorobenzoyl)urea

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H8Cl2F8N2O3
CAS Number:
Molecular Weight:
511.15
Beilstein:
8398291
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

lufenuron

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

FC(C(F)(F)F)C(F)(F)Oc1cc(Cl)c(NC(=O)NC(=O)c2c(F)cccc2F)cc1Cl

InChI

1S/C17H8Cl2F8N2O3/c18-6-5-11(32-17(26,27)14(22)16(23,24)25)7(19)4-10(6)28-15(31)29-13(30)12-8(20)2-1-3-9(12)21/h1-5,14H,(H2,28,29,30,31)

InChI key

PWPJGUXAGUPAHP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Lufenuron EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

>338.0 °F - (External MSDS)

Flash Point(C)

> 170 °C - (External MSDS)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Martín Breijo et al.
Veterinary parasitology, 181(2-4), 341-344 (2011-05-20)
The aim of this work was to evaluate the potential of lufenuron, a benzylphenylurea with ability to interfere with the formation of insect exoskeleton, as a therapeutic drug for larval echinococcosis (hydatid disease). For this purpose lufenuron, alone or in
V Navarro-Llopis et al.
Journal of economic entomology, 104(5), 1647-1655 (2011-11-10)
During 2008 and 2009, the efficacy of the combination of two Mediterranean fruit fly, Ceratitis capitata (Wiedemann) (Diptera: Tephritidae), control techniques, sterile insect technique (SIT) and a chemosterilant bait station system (Adress), was tested in three crops: citrus (Citrus spp.)
Patricia López-Mancisidor et al.
Environmental toxicology and chemistry, 27(6), 1317-1331 (2008-05-10)
Outdoor experimental ditches were used to evaluate the influence of untreated refuges on the recovery of zooplankton communities following treatment with the fast-dissipating insecticide lufenuron. Each experimental ditch was divided into three sections of the same surface area. The treatments
Vicente Navarro-Llopis et al.
Pest management science, 66(5), 511-519 (2010-01-27)
The chemosterilisation technique has been demonstrated to reduce the population and fruit damage of the Mediterranean fruit fly, Ceratitis capitata (Wiedemann), in citrus orchards. Field trials showed efficacy by reducing the fruit fly population, which was progressively achieved by continuous
Theo C M Brock et al.
Environmental toxicology and chemistry, 29(9), 1994-2008 (2010-09-08)
Under typical agricultural use of an insecticide, it is likely that only part of an edge-of-field drainage ditch will be directly contaminated by spray drift. The response, including recovery, of aquatic macroinvertebrates in sprayed ditch sections may be affected by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service